Skip to Content
Merck
All Photos(1)

Key Documents

30-3350

Sigma-Aldrich

p-Toluenesulfonic acid monohydrate

SAJ special grade, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4SO3H · H2O
CAS Number:
Molecular Weight:
190.22
Beilstein:
3568023
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

grade

SAJ special grade

vapor density

5.9 (vs air)

Assay

≥99.0%

form

solid

availability

available only in Japan

mp

103-106 °C (lit.)

SMILES string

[H]O[H].Cc1ccc(cc1)S(O)(=O)=O

InChI

1S/C7H8O3S.H2O/c1-6-2-4-7(5-3-6)11(8,9)10;/h2-5H,1H3,(H,8,9,10);1H2

InChI key

KJIFKLIQANRMOU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Shanmugam et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 95, 177-183 (2012-05-29)
Bulk crystals of newly identified organic nonlinear optical material 4-methylanilinium p-toluenesulfonate (PMPT) were grown by slow evaporation solution growth method using ethanol as a solvent. It crystallizes in monoclinic system with a noncentrosymmetric space group P2(1). The formation of the
Mild C-H halogenation of anilides and the isolation of an unusual palladium(I)-palladium(II) species.
Robin B Bedford et al.
Angewandte Chemie (International ed. in English), 50(24), 5524-5527 (2011-05-14)
Hans H Adomat et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 902, 84-95 (2012-07-24)
Androgens are key mediators of prostate development and function, a role that extends to the development of prostate diseases such as benign prostatic hyperplasia (BPH) and prostate cancer. In prostate, DHT is the major androgen and reduction and glucuronidation are
Pei Liu et al.
Organic & biomolecular chemistry, 10(24), 4696-4698 (2012-05-18)
A convenient one-pot Mannich-type-cyclization-oxidation tandem process has been developed for the synthesis of 1,3,5-trisubstituted pyrazoles derivatives from aldehydes, hydrazines and alkynes using p-toluenesulfonic acid monohydrate (PTSA) as a multifunctional catalyst. This method provides a flexible and rapid route to 1,3,5-trisubstituted
Jhonny Azuaje et al.
ACS combinatorial science, 13(1), 89-95 (2011-01-21)
We document here the use of polymer-supported p-toluenesulfonic acid as a highly effective, robust, economical and eco-friendly isocyanide scavenger. The herein described strategy circumvent the intense and repulsive odor of volatile isocyanides, enabling simplified and odorless workup and purifications. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service