Skip to Content
Merck
All Photos(1)

Key Documents

28-5760

Sigma-Aldrich

Sulfanilic acid

99.0-100.5%, suitable for determination of nitroxide

Synonym(s):

4-Aminobenzenesulfonic acid, Aniline-4-sulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(H2N)C6H4SO3H
CAS Number:
Molecular Weight:
173.19
Beilstein:
908765
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

Assay

99.0-100.5%

form

powder

availability

available only in Japan

mp

>300 °C (lit.)

solubility

water: soluble 12.51 g/L at 20 °C

density

1.4862 g/cm3 at 20 °C

suitability

suitable for determination of nitroxide

SMILES string

Nc1ccc(cc1)S(O)(=O)=O

InChI

1S/C6H7NO3S/c7-5-1-3-6(4-2-5)11(8,9)10/h1-4H,7H2,(H,8,9,10)

InChI key

HVBSAKJJOYLTQU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Han Ming Gan et al.
Microbiology (Reading, England), 158(Pt 8), 1933-1941 (2012-05-23)
The gene coding for the oxygenase component, sadA, of 4-aminobenzenesulfonate (4-ABS) 3,4-dioxygenase in Hydrogenophaga sp. PBC was previously identified via transposon mutagenesis. Expression of wild-type sadA in trans restored the ability of the sadA mutant to grow on 4-ABS. The
Han Ming Gan et al.
Chemosphere, 82(4), 507-513 (2010-11-26)
A co-culture consisting of Hydrogenophaga sp. PBC and Ralstonia sp. PBA, isolated from textile wastewater treatment plant could tolerate up to 100 mM 4-aminobenzenesulfonate (4-ABS) and utilize it as sole carbon, nitrogen and sulfur source under aerobic condition. The biodegradation
Jinsong Zhang et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 63(11), 2752-2758 (2011-11-05)
4-Aminobenzenesulfonate (4-ABS), an aromatic amine and recalcitrant toxic pollutant, is widely used in the dye and pharmaceutical industry. Pannonibactersp. W1 is a specialized microbial strain which can efficiently degrade 4-ABS. This study shows the feasibility of using the specialized strain
Yadong Sun et al.
Organic letters, 15(7), 1598-1601 (2013-03-19)
An efficient and convenient method was developed for the formation of 2-substituted benzothiazoles via a copper-catalyzed condensation of 2-aminobenzenethiols with nitriles. The developed method is applicable to a wide range of nitriles containing different functional groups furnishing excellent yields of
Nam Hoon Kim et al.
Nano letters, 10(10), 4181-4185 (2010-09-25)
A bifunctional adenosine-sensitive double-stranded DNA aptamer was used to create and control a surface-enhanced Raman spectroscopy (SERS) hot spot between a bulk Au surface and a gold nanoparticle (Au NP) attached to the aptamer via a biotin-avidin linkage. The Au

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service