Skip to Content
Merck
All Photos(1)

Documents

28-5031

Sigma-Aldrich

Stearic acid

SAJ first grade, ≥90.0%, powder

Synonym(s):

1-Heptadecanecarboxylic acid, C18:0, Cetylacetic acid, NSC 25956, NSC 261168, Octadecanoic acid, Stearophanic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)16COOH
CAS Number:
Molecular Weight:
284.48
Beilstein:
608585
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

SAJ first grade

vapor pressure

1 mmHg ( 173.7 °C)

Assay

≥90.0%

form

powder

availability

available only in Japan

bp

361 °C (lit.)

mp

67-72 °C (lit.)

density

0.845 g/cm3

functional group

carboxylic acid

storage temp.

room temp

SMILES string

CCCCCCCCCCCCCCCCCC(O)=O

InChI

1S/C18H36O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h2-17H2,1H3,(H,19,20)

InChI key

QIQXTHQIDYTFRH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ulla Hoppu et al.
Journal of pediatric gastroenterology and nutrition, 41(3), 335-338 (2005-09-01)
Breast milk fatty acids may have immunomodulatory properties related to the development of atopic disease. The aim of this study was to assess the impact of the breast milk fatty acid composition on the development of atopic dermatitis (AD) in
Li Zeng et al.
PloS one, 3(6), e2329-e2329 (2008-06-05)
DNA damage triggers a network of signaling events that leads to cell cycle arrest or apoptosis. This DNA damage response acts as a mechanism to prevent cancer development. It has been reported that fatty acids (FAs) synthesis is increased in
Piret Arukuusk et al.
Biochimica et biophysica acta, 1828(5), 1365-1373 (2013-01-30)
Harnessing of a branched structure is a novel approach in the design of cell-penetrating peptides and it has provided highly efficient transfection reagents for intracellular delivery of nucleic acids. The new stearylated TP10 analogs, NickFects, condense plasmid DNA, splice correcting
T A Doran et al.
American journal of obstetrics and gynecology, 133(3), 302-307 (1979-02-01)
Lecithin/sphingomyelin (L/S) ratio, creatinine, percentage of lipid-positive cells, palmitic acid, palmitic acid/stearic acid (P/S) ratio, and total cortisol were analyzed as tests for fetal maturity and fetal pulmonary maturity in 164 samples of amniotic fluid from 121 patients. Fifty samples
Qian Wan et al.
Nanoscale, 5(2), 744-752 (2012-12-12)
As a versatile gene vector, minicircle DNA (mcDNA) has a great potential for gene therapy. However, some serious challenges remain, such as to effectively deliver mcDNA into targeted cells/tissues and to non-invasively monitor the delivery of the mcDNA. Superparamagnetic iron

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service