Skip to Content
Merck
All Photos(1)

Key Documents

28-4760

Sigma-Aldrich

Sorbic acid

SAJ first grade, ≥98.5%

Synonym(s):

α-trans-γ-trans-Sorbic acid, (E,E)-1,3-Pentadiene-1-carboxylic acid, 2,4-Hexadienoic acid, 2E,4E-Hexadienoic acid, trans,trans-2,4-Hexadienoic acid, C6:2n-2,4, NSC 35405, NSC 49103, NSC 50268

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHCH=CHCOOH
CAS Number:
Molecular Weight:
112.13
Beilstein:
1098547
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
Pricing and availability is not currently available.

grade

SAJ first grade

vapor pressure

0.01 mmHg ( 20 °C)

Assay

≥98.5%

form

crystalline

availability

available only in Japan

mp

132-135 °C (lit.)

density

1.2 g/cm3 at 20 °C

antibiotic activity spectrum

fungi

Mode of action

enzyme | inhibits

SMILES string

C\C=C\C=C\C(O)=O

InChI

1S/C6H8O2/c1-2-3-4-5-6(7)8/h2-5H,1H3,(H,7,8)/b3-2+,5-4+

InChI key

WSWCOQWTEOXDQX-MQQKCMAXSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

260.6 °F - closed cup

Flash Point(C)

127 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G D Saraiva et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 137, 1409-1416 (2014-10-14)
This work reports a temperature-dependent vibrational spectroscopic study of the sorbic acid (C6H8O2), as well as the mode assignment at ambient conditions, based on the density functional theory. Temperature-dependent vibrational properties have been performed in polycrystalline sorbic acid through both
A Vermeulen et al.
Food microbiology, 32(2), 389-396 (2012-09-19)
The microbial stability of intermediate moisture foods (IMF) is linked with the possible growth of osmophilic yeast and xerophilic moulds. As most of these products have a long shelf life the assessment of the microbial stability is often an important
Robert Mueller et al.
Langmuir : the ACS journal of surfaces and colloids, 28(27), 10296-10303 (2012-06-15)
Carbon molecular sieve (CMS) membranes are promising materials for energy efficient separations of light gases. In this work, we report a detailed microscopic study of carbon dioxide and methane self-diffusion in three CMS membrane derived from 6FDA/BPDA(1:1)-DAM and Matrimid polymers.
Phercyles Veiga-Santos et al.
Acta tropica, 125(3), 349-356 (2012-12-12)
The currently available treatments for Chagas disease show limited therapeutic potential and are associated with serious side effects. Our group has been attempting to find alternative drugs isolated from natural products as a potential source of pharmacological agents against Trypanosoma
Heidy M W den Besten et al.
Applied and environmental microbiology, 78(23), 8477-8480 (2012-09-25)
Population heterogeneity complicates the predictability of the outgrowth kinetics of individual spores. Flow cytometry sorting and monitoring of the germination and outgrowth of single dormant spores allowed the quantification of acid-induced spore population heterogeneity at pH 5.5 and in the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service