Skip to Content
Merck
All Photos(1)

Key Documents

28-4730

Sigma-Aldrich

Somogyi′s copper reagent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
grade:
SAJ special grade
form:
liquid
Pricing and availability is not currently available.

grade

SAJ special grade

form

liquid

availability

available only in Japan

storage temp.

15-25°C

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service