Skip to Content
Merck
All Photos(1)

Documents

28-3660

Sigma-Aldrich

Sodium peroxide

SAJ first grade, ≥90.0%

Synonym(s):

Sodium superoxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Na2O2
CAS Number:
Molecular Weight:
77.98
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:

grade

SAJ first grade

Assay

≥90.0%

form

powder

reaction suitability

reagent type: oxidant

availability

available only in Japan

mp

460 °C (dec.) (lit.)

SMILES string

[Na+].[Na+].[O-][O-]

InChI

1S/2Na.O2/c;;1-2/q2*+1;-2

InChI key

PFUVRDFDKPNGAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame over circleCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Ox. Sol. 1 - Skin Corr. 1A

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christine Sedgley
Journal of the history of dentistry, 52(2), 61-65 (2004-08-06)
The concept of the germ theory of disease combined with the development of dentistry during the latter half of the 19th century had a direct effect on the practice of endodontics. The significance of root canal irrigation to endodontics strengthened
K J Sreeram et al.
Journal of environmental monitoring : JEM, 3(5), 526-530 (2001-11-07)
The processing of chromite ore is associated with the generation of large quantities of solid wastes containing chromium, which have been disposed of as landfill for many years. The mobilization and operational speciation of chromium contained in soils contaminated with
N C Verma et al.
Journal of environmental pathology, toxicology and oncology : official organ of the International Society for Environmental Toxicology and Cancer, 20(1), 1-7 (2001-02-24)
Saccharomyces cerevisiae shows altered radiation response under various stress conditions, such as nutrition depletion, nitrogen starvation, osmotic shock, heat shock, and mild chemical treatments. In general, the cells show higher levels of UV or gamma radiation resistance under the stress.
[Rapid ultraviolet-absorption photometric determination of the protein content of aqueous extracts in the microgram area using Na2O2].
B Löhrke
Zeitschrift fur medizinische Laboratoriumsdiagnostik, 27(2), 92-95 (1986-01-01)
S Kesten et al.
Annals of allergy, asthma & immunology : official publication of the American College of Allergy, Asthma, & Immunology, 74(3), 269-273 (1995-03-01)
Atmospheric pollution has been proposed as one of the possible factors responsible for increases in asthma mortality and morbidity. We sought to examine whether we could demonstrate a relationship between emergency room visits for asthma and alterations in environmental conditions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service