28-2650
Sodium ethoxide
SAJ first grade, ≥95.0%
Synonym(s):
Sodium ethylate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3CH2ONa
CAS Number:
Molecular Weight:
68.05
Beilstein:
3593646
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
grade
SAJ first grade
vapor density
1.6 (vs air)
vapor pressure
<0.1 mmHg ( 20 °C)
Assay
≥95.0%
form
solid
availability
available only in Japan
SMILES string
[Na+].CC[O-]
InChI
1S/C2H5O.Na/c1-2-3;/h2H2,1H3;/q-1;+1
InChI key
QDRKDTQENPPHOJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Sol. 1 - Self-heat. 1 - Skin Corr. 1A
Supplementary Hazards
Storage Class Code
4.2 - Pyrophoric and self-heating hazardous materials
WGK
WGK 1
Flash Point(F)
86.0 °F - closed cup
Flash Point(C)
30 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Zhao Cai et al.
Applied immunohistochemistry & molecular morphology : AIMM, 13(3), 292-294 (2005-08-06)
The authors describe a simple and cost-effective method of immunostaining semithin epoxy resin sections of peripheral nerve for light microscopy with antibodies to myelin protein zero, peripheral myelin protein 22, myelin basic protein, and neurofilament protein 200 using a combined
Franklin R Tay et al.
Journal of endodontics, 31(8), 593-598 (2005-07-27)
Polycaprolactone, a thermoplastic aliphatic polyester, is reportedly susceptible to both alkaline and enzymatic hydrolyzes. This screening study examined the susceptibility of Resilon, a polycaprolactone-based root filling composite, to alkaline hydrolysis. There were 15-mm diameter disks of Resilon and Obtura gutta-percha
N C Luu et al.
Chemical research in toxicology, 13(7), 610-615 (2000-07-18)
Glutathione conjugate formation plays important roles in the detoxification and bioactivation of xenobiotics. A range of nephrotoxic haloalkenes undergo bioactivation that involves glutathione and cysteine S-conjugate formation. The cysteine S-conjugates thus formed may undergo cysteine conjugate beta-lyase-catalyzed biotransformation to form
Jung Mi Kwon et al.
Journal of hazardous materials, 134(1-3), 230-236 (2005-12-20)
A photocatalyst was prepared by attaching TiO(2) powder (diameter, 50nm) in the sol state to fluidizing spherical ceramic carriers using a silicon binder. A high initial photocatalytic activity and strong attachment was obtained at a sintering temperature of 500 degrees
S H Brorson et al.
Micron (Oxford, England : 1993), 26(4), 301-310 (1995-01-01)
The purpose of this investigation was to explain why deplasticizing of epoxy sections gives higher immunogold labeling than non-deplasticizing. The methods used were the following: (1) Comparison of the ratio of immunogold labeling of deplasticized and non-deplasticized sections with gold
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service