Skip to Content
Merck
All Photos(1)

Key Documents

28-2250

Sigma-Aldrich

Sodium chlorate

SAJ first grade, ≥98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaClO3
CAS Number:
Molecular Weight:
106.44
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Assay:
≥98.0%
grade:
SAJ first grade
form:
solid
Pricing and availability is not currently available.

grade

SAJ first grade

Assay

≥98.0%

form

solid

availability

available only in Japan

mp

248-261 °C (lit.)

storage temp.

15-25°C

SMILES string

[Na+].[O-]Cl(=O)=O

InChI

1S/ClHO3.Na/c2-1(3)4;/h(H,2,3,4);/q;+1/p-1

InChI key

YZHUMGUJCQRKBT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

sulfation biosynthesis inhibitor

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Ox. Sol. 1

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ha Bich Trinh et al.
Journal of hazardous materials, 379, 120772-120772 (2019-06-30)
Total recycling of all valuable metals such as PGMs, Ce, Al and Mg from the spent automobile catalysts has been explored. The alkali roasting was performed under NaOH, 0.5-3.0 g; temperature, 300-800 °C; and time, 10-60 min. The phase transformation from cordierite to
Ludmila Abezgauz et al.
Journal of colloid and interface science, 342(1), 83-92 (2009-11-27)
Controlling the morphological characteristics of micellar solutions is important for surfactant performance and for achieving desired properties. In this work we study how monovalent anions of the lyotropic series affect micellization, micellar transitions, and micellar growth of the cationic surfactant
Fei Wang et al.
Chemosphere, 89(8), 1009-1014 (2012-08-18)
Understanding the interaction of perfluorochemicals, persistent pollutants with known human health effects, with mineral compounds in surface water and groundwater environments is essential to determining their fate and transport. Kinetic experiments showed that adsorption equilibrium can be achieved within 48
Stoyan I Karakashev et al.
Langmuir : the ACS journal of surfaces and colloids, 24(20), 11587-11591 (2008-09-12)
We report the effects of ions on rupture and lifetime of aqueous foam films formed from sodium chloride (NaCl), lithium chloride (LiCl), sodium acetate (NaAc), and sodium chlorate (NaClO 3) using microinterferometry. In the case of NaCl and LiCl, the
Riia Plihtari et al.
Atherosclerosis, 217(2), 401-406 (2011-05-17)
The extracellular pH is locally decreased in advanced atherosclerotic lesions, particularly in lipid-rich areas of the lesions. Since accumulation of LDL-derived cholesterol and formation of foam cells are key processes in atherogenesis, we tested here the effects of acidic pH

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service