Skip to Content
Merck
All Photos(1)

Key Documents

24-4540

Sigma-Aldrich

Potassium hexacyanoferrate(III)

SAJ first grade, ≥98.0%

Synonym(s):

Potassium ferricyanide, Red prussiate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K3Fe(CN)6
CAS Number:
Molecular Weight:
329.24
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
SAJ first grade
form:
crystalline
Pricing and availability is not currently available.

grade

SAJ first grade

Assay

≥98.0%

form

crystalline

reaction suitability

reagent type: catalyst
core: iron

availability

available only in Japan

pH

6.0-9.0 (25 °C, 329 g/L)

SMILES string

[K+].[K+].[K+].N#C[Fe-3](C#N)(C#N)(C#N)(C#N)C#N

InChI

1S/6CN.Fe.3K/c6*1-2;;;;/q;;;;;;-3;3*+1

InChI key

MIMJFNVDBPUTPB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2

Supplementary Hazards

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daehee Kim et al.
ChemSusChem, 5(6), 1086-1091 (2012-05-10)
To scale-up microbial fuel cells (MFCs), installing multiple unit cells in a common reactor has been proposed; however, there has been a serious potential drop when connecting unit cells in series. To determine the source of the loss, a basic
Liju Yang et al.
Biomedical microdevices, 13(2), 279-289 (2010-11-26)
Microlithographically fabricated interdigitated microsensor electrodes (IMEs) were cleaned, surface activated, chemically functionalized (amine) and derivatized with an Acrloyl-PEG-NHS to receive a spun-applied monomer cocktail of UV polymerizable monomer. IMEs were 2050.5, 1550.5, 1050.5 and 0550.5 possessing lines and spaces that
Hideo Takakusa et al.
Drug metabolism and disposition: the biological fate of chemicals, 39(6), 1022-1030 (2011-03-03)
Lapatinib, an oral breast cancer drug, has recently been reported to be a mechanism-based inactivator of cytochrome P450 (P450) 3A4 and also an idiosyncratic hepatotoxicant. It was suggested that formation of a reactive quinoneimine metabolite was involved in mechanism-based inactivation
Lu Han et al.
Journal of pharmaceutical and biomedical analysis, 58, 141-145 (2011-10-15)
In this paper, a novel chemiluminescence (CL) system, 2-phenyl-4, 5-di (2-furyl) imidazole (PDFI)-potassium ferricyanide, for the determination of terbutaline sulfate was described. The method was based on enhancement of CL emission of PDFI-potassium ferricyanide system in the presence of terbutaline
Anna Poladyan et al.
Current microbiology, 66(1), 49-55 (2012-10-12)
Glycerol and glucose fermentation redox routes by Escherichia coli and their regulation by oxidizing and reducing reagents were investigated at different pHs. Cell growth was followed by decrease of pH and redox potential (E ( h )). During glycerol utilization

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service