Skip to Content
Merck
All Photos(1)

Documents

24-1405

Sigma-Aldrich

Phenol

≥99.0%

Synonym(s):

Hydroxybenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5OH
CAS Number:
Molecular Weight:
94.11
Beilstein:
969616
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

vapor density

3.24 (vs air)

vapor pressure

0.09 psi ( 55 °C)
0.36 mmHg ( 20 °C)

Assay

≥99.0%

form

solid

autoignition temp.

1319 °F

expl. lim.

8.6 %

availability

available only in Japan

pH

~5 (20 °C, 50 g/L)

bp

182 °C (lit.)

mp

40-42 °C (lit.)

density

1.071 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

Oc1ccccc1

InChI

1S/C6H6O/c7-6-4-2-1-3-5-6/h1-5,7H

InChI key

ISWSIDIOOBJBQZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Suitability

for biological purposes

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - STOT RE 2

Target Organs

Nervous system,Kidney,Liver,Skin

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

174.2 °F

Flash Point(C)

79 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thitiporn Anunthawan et al.
Biochimica et biophysica acta, 1848(6), 1352-1358 (2015-03-15)
We investigated the mechanisms of two tryptophan-rich antibacterial peptides (KT2 and RT2) obtained in a previous optimization screen for increased killing of both Gram-negative and Gram-positive bacteria pathogens. At their minimal inhibitory concentrations (MICs), these peptides completely killed cells of
Rita A Busuttil et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 20(10), 2761-2772 (2014-03-25)
Gene-expression profiling has revolutionized the way we think about cancer and confers the ability to observe the synchronous expression of thousands of genes. The use of putative genome-level expression profiles has allowed biologists to observe the complex interactions of genes
Mark J Lee et al.
PLoS pathogens, 11(10), e1005187-e1005187 (2015-10-23)
Of the over 250 Aspergillus species, Aspergillus fumigatus accounts for up to 80% of invasive human infections. A. fumigatus produces galactosaminogalactan (GAG), an exopolysaccharide composed of galactose and N-acetyl-galactosamine (GalNAc) that mediates adherence and is required for full virulence. Less
Yukari Yamane-Sando et al.
MicrobiologyOpen, 3(2), 196-212 (2014-02-11)
Sphingolipids are a family of eukaryotic lipids biosynthesized from sphingoid long-chain bases (LCBs). Sphingolipids are an essential class of lipids, as their depletion results in cell death. However, acute LCB supplementation is also toxic; thus, proper cellular LCB levels should
N C Chapman et al.
Heredity, 114(6), 584-592 (2015-01-15)
Asexual reproduction via thelytokous parthenogenesis is widespread in the Hymenoptera, but its genetic underpinnings have been described only twice. In the wasp Lysiphlebus fabarum and the Cape honey bee Apis mellifera capensis the origin of thelytoky have each been traced

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service