Skip to Content
Merck
All Photos(1)

Key Documents

24-0170

Sigma-Aldrich

Palmitic acid

SAJ first grade, ≥95.0%

Synonym(s):

1-Pentadecanecarboxylic acid, C16:0, Cetylic acid, Hexadecanoic acid, NSC 5030, PamOH

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)14COOH
CAS Number:
Molecular Weight:
256.42
Beilstein:
607489
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

SAJ first grade

vapor pressure

10 mmHg ( 210 °C)

Assay

≥95.0%

form

flakes

availability

available only in Japan

bp

271.5 °C/100 mmHg (lit.)

mp

61-62.5 °C (lit.)

solubility

water: 0.00005 g/L at 20 °C

density

0.852 g/mL at 25 °C (lit.)

functional group

carboxylic acid

storage temp.

room temp

SMILES string

CCCCCCCCCCCCCCCC(O)=O

InChI

1S/C16H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h2-15H2,1H3,(H,17,18)

InChI key

IPCSVZSSVZVIGE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

235.4 °F

Flash Point(C)

113 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jonas Sieber et al.
The American journal of pathology, 183(3), 735-744 (2013-07-23)
Type 2 diabetes mellitus is characterized by dyslipidemia with elevated free fatty acids (FFAs). Loss of podocytes is a hallmark of diabetic nephropathy, and podocytes are highly susceptible to saturated FFAs but not to protective, monounsaturated FFAs. We report that
B Vessby et al.
Diabetologia, 37(10), 1044-1050 (1994-10-01)
Recent data indicate that peripheral insulin sensitivity may be influenced by dietary fat quality and skeletal muscle phospholipid fatty acid composition. During a health survey of 70-year-old men insulin sensitivity was measured by the euglycaemic hyperinsulinaemic clamp technique and the
Hjalti Kristinsson et al.
Endocrinology, 154(11), 4078-4088 (2013-09-17)
Free fatty acids (FFAs) have pleiotropic effects on the pancreatic β-cell. Although acute exposure to FFAs stimulates glucose-stimulated insulin secretion (GSIS), prolonged exposure impairs GSIS and causes apoptosis. FFAs exert their effects both via intracellular metabolism and interaction with the
Mathieu Blanc et al.
Biochemical Society transactions, 41(1), 84-88 (2013-01-30)
S-Palmitoylation, the only reversible post-translational lipid modification, confers unique biochemical and functional properties to proteins. Although it has long been known that viral proteins are palmitoylated, recent studies reveal that this modification plays a critical role for pathogens of all
Sanja Blaskovic et al.
The FEBS journal, 280(12), 2766-2774 (2013-04-05)
S-palmitoylation is post-translational modification, which consists in the addition of a C16 acyl chain to cytosolic cysteines and which is unique amongst lipid modifications in that it is reversible. It can thus, like phosphorylation or ubiquitination, act as a switch.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service