Skip to Content
Merck
All Photos(1)

Key Documents

21-2050

Sigma-Aldrich

Nitrobenzene

SAJ first grade, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5NO2
CAS Number:
Molecular Weight:
123.11
Beilstein:
507540
MDL number:
UNSPSC Code:
12191501
PubChem Substance ID:
grade:
SAJ first grade
bp:
210-211 °C (lit.)
vapor pressure:
0.15 mmHg ( 20 °C)
50 mmHg ( 120 °C)

grade

SAJ first grade

vapor density

4.2 (vs air)

vapor pressure

0.15 mmHg ( 20 °C)
50 mmHg ( 120 °C)

Assay

≥99.0%

form

liquid

autoignition temp.

899 °F

expl. lim.

40 %

availability

available only in Japan

dilution

(for analytical testing)

refractive index

n20/D 1.551 (lit.)

pH

8.0-8.5 (20 °C, 1 g/L)

bp

210-211 °C (lit.)

mp

5-6 °C (lit.)

density

1.196 g/mL at 25 °C (lit.)

SMILES string

[O-][N+](=O)c1ccccc1

InChI

1S/C6H5NO2/c8-7(9)6-4-2-1-3-5-6/h1-5H

InChI key

LQNUZADURLCDLV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Carc. 2 - Repr. 1B - STOT RE 1 Inhalation

Target Organs

Blood

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

190.4 °F

Flash Point(C)

88 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yufan Zhang et al.
Analytica chimica acta, 752, 45-52 (2012-10-30)
Novel Pt nanoparticles (PN) ensemble on macroporous carbon (MPC) hybrid nanocomposites (PNMPC) were prepared through a rapidly and simple one-step microwave-assisted heating procedure. The obtained PNMPC was characterized by transmission electron microscopy (TEM), X-ray photoelectron spectroscopy (XPS), thermogravimetric analysis (TGA)
Iwona Cybulska et al.
Bioresource technology, 118, 30-36 (2012-06-15)
Lignin extracted from prairie cordgrass, switchgrass, and corn stover (using ethyl acetate-ethanol-water organosolv pretreatment) was analyzed and characterized using several methods. These methods included analysis of purity (by determination of Klason lignin, carbohydrate, and ash contents), solubility (with several organic
Jingang Huang et al.
Water research, 46(14), 4361-4370 (2012-06-19)
Sulfate is frequently found in wastewaters that contain nitrobenzene. To reveal the effect of sulfate on the reductive transformation of nitrobenzene to aniline--with acetate or propionate as potential electron donors in anaerobic systems--an acetate series (R1-R5) and a propionate series
Krzysztof Zasada et al.
Toxicology and industrial health, 31(12), 1195-1201 (2013-06-01)
Nitrobenzene is a carcinogen, which induces-among others-thyroid tumors. Melatonin is an effective antioxidant, whereas some antioxidative effects of propylthiouracil (PTU; an antithyroid medication used for the treatment of thyrotoxicosis) were also found. The aim of the study was to compare
Jiping Jiang et al.
Journal of hazardous materials, 227-228, 280-291 (2012-06-06)
This paper presents a generic framework and decision tools of real-time risk assessment on Emergency Environmental Decision Support System for response to chemical spills in river basin. The generic "4-step-3-model" framework is able to delineate the warning area and the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service