Skip to Content
Merck
All Photos(1)

Key Documents

21-2040

Sigma-Aldrich

Nitrobenzene

JIS special grade, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5NO2
CAS Number:
Molecular Weight:
123.11
Beilstein:
507540
MDL number:
UNSPSC Code:
12191501
PubChem Substance ID:
grade:
JIS special grade
bp:
210-211 °C (lit.)
vapor pressure:
0.15 mmHg ( 20 °C)
50 mmHg ( 120 °C)
Pricing and availability is not currently available.

grade

JIS special grade

vapor density

4.2 (vs air)

vapor pressure

0.15 mmHg ( 20 °C)
50 mmHg ( 120 °C)

Assay

≥99.5%

form

liquid

autoignition temp.

899 °F

expl. lim.

40 %

availability

available only in Japan

dilution

(for analytical testing)

refractive index

n20/D 1.551 (lit.)

pH

8.0-8.5 (20 °C, 1 g/L)

bp

210-211 °C (lit.)

mp

5-6 °C (lit.)

density

1.196 g/mL at 25 °C (lit.)

SMILES string

[O-][N+](=O)c1ccccc1

InChI

1S/C6H5NO2/c8-7(9)6-4-2-1-3-5-6/h1-5H

InChI key

LQNUZADURLCDLV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Carc. 2 - Repr. 1B - STOT RE 1 Inhalation

Target Organs

Blood

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

190.4 °F

Flash Point(C)

88 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Emmanuel et al.
Journal of hazardous materials, 279, 117-124 (2014-07-23)
The present study involves a green synthesis of gold nanoparticles (Au-NPs) using Acacia nilotica twig bark extract at room temperature and trace level detection of one of the hazardous materials, viz. nitrobenzene (NB) that causes Methemoglobinaemia. The synthesis protocol demonstrates
Emilia M Reyes-Gallardo et al.
Journal of chromatography. A, 1271(1), 50-55 (2012-12-15)
In this article, the combination of commercial polymeric microparticles (OASIS MCX) and cobalt ferrite nanoparticles is evaluated in dispersive micro-solid phase extraction (D-μSPE) for the determination of six nitroaromatic hydrocarbons in water. The high affinity of the polymeric material toward
Jie Xie et al.
Journal of hazardous materials, 231-232, 57-63 (2012-07-10)
The aim of this study was to examine the adsorption capability and mechanism of hexadecyltrimethylammonium modified zeolite, which was synthesized from coal fly ash, for the removal of ionizable phenolic compounds (phenol, p-chlorophenol and bisphenol A, with different pK(a)) and
Jingang Huang et al.
Water research, 46(14), 4361-4370 (2012-06-19)
Sulfate is frequently found in wastewaters that contain nitrobenzene. To reveal the effect of sulfate on the reductive transformation of nitrobenzene to aniline--with acetate or propionate as potential electron donors in anaerobic systems--an acetate series (R1-R5) and a propionate series
Iwona Cybulska et al.
Bioresource technology, 118, 30-36 (2012-06-15)
Lignin extracted from prairie cordgrass, switchgrass, and corn stover (using ethyl acetate-ethanol-water organosolv pretreatment) was analyzed and characterized using several methods. These methods included analysis of purity (by determination of Klason lignin, carbohydrate, and ash contents), solubility (with several organic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service