Skip to Content
Merck
All Photos(1)

Key Documents

21-1240

Sigma-Aldrich

Nickel(II) oxide

CP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NiO
CAS Number:
Molecular Weight:
74.69
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
grade:
CP
form:
granular

grade

CP

form

granular

availability

available only in Japan

density

6.67 g/mL at 25 °C (lit.)

SMILES string

[Ni]=O

InChI

1S/Ni.O

InChI key

GNRSAWUEBMWBQH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Carc. 1A Inhalation - Skin Sens. 1 - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elizabeth A Gibson et al.
Physical chemistry chemical physics : PCCP, 15(7), 2411-2420 (2013-01-10)
Photoactive NiO electrodes for cathodic dye-sensitised solar cells (p-DSCs) have been prepared with thicknesses ranging between 0.4 and 3.0 μm by spray-depositing pre-formed NiO nanoparticles on fluorine-doped tin oxide (FTO) coated glass substrates. The larger thicknesses were obtained in sequential
Salvador B Muñoz et al.
Inorganic chemistry, 51(23), 12660-12668 (2012-11-13)
The synthesis and characterization of new tris(carbene)borate ligand precursors containing substituted benzimidazol-2-ylidene and 1,3,4-triazol-2-ylidene donor groups, as well as a new tris(imidazol-2-ylidene)borate ligand precursor are reported. The relative donor strengths of the tris(carbene)borate ligands have been evaluated by the position
Neda Mazinanian et al.
Regulatory toxicology and pharmacology : RTP, 65(1), 135-146 (2012-11-13)
Differences in surface oxide characteristics and extent of nickel release have been investigated in two thoroughly characterized micron-sized (mainly <4 μm) nickel metal powders and a nickel oxide bulk powder when immersed in two different synthetic fluids, artificial sweat (ASW-pH
Huaixiang Li et al.
Biosensors & bioelectronics, 47, 225-230 (2013-04-16)
A novel photoelectrochemical hydrogen peroxide (H2O2) sensor was constructed with platinum (Pt) and nickel hydroxyl-oxide (NiOOH) double layers modified n-silicon electrode (NiOOH/Pt/n-n(+)-Si). About 40nm Pt layer and about 100nm Ni layer were successively coated on the front surface of n-n(+)-Si
Genqiang Zhang et al.
Nanoscale, 5(3), 877-881 (2012-12-15)
One-dimensional hierarchical hollow nanostructures composed of NiO nanosheets are successfully synthesized through a facile carbon nanofiber directed solution method followed by a simple thermal annealing treatment. With the advantages of high electro-active surface area, carbon nanofiber supported robust structure and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service