Skip to Content
Merck
All Photos(1)

Key Documents

19-1730

Sigma-Aldrich

Mercury(II) nitrate hydrate

JIS special grade, ≥95.0%

Synonym(s):

Mercury nitrate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Hg(NO3)2
Molecular Weight:
324.60 (anhydrous basis)
UNSPSC Code:
12352300
grade:
JIS special grade
form:
solid
Pricing and availability is not currently available.

grade

JIS special grade

Assay

≥95.0%

form

solid

availability

available only in Japan

storage temp.

15-25°C

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service