Skip to Content
Merck
All Photos(1)

Key Documents

19-0670

Sigma-Aldrich

DL-Malic acid

SAJ first grade, ≥99.0%

Synonym(s):

(±)-2-Hydroxysuccinic acid, DL-Hydroxybutanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH2CH(OH)CO2H
CAS Number:
Molecular Weight:
134.09
Beilstein:
1723539
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
grade:
SAJ first grade
vapor pressure:
<0.1 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

SAJ first grade

vapor density

4.6 (vs air)

vapor pressure

<0.1 mmHg ( 20 °C)

Assay

≥99.0%

form

solid

autoignition temp.

644 °F

availability

available only in Japan

dilution

(for analytical testing)

mp

131-133 °C (lit.)

SMILES string

OC(CC(O)=O)C(O)=O

InChI

1S/C4H6O5/c5-2(4(8)9)1-3(6)7/h2,5H,1H2,(H,6,7)(H,8,9)

InChI key

BJEPYKJPYRNKOW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

397.4 °F

Flash Point(C)

203 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaojie Liu et al.
Journal of chromatography. A, 1532, 198-207 (2017-12-13)
Natural deep eutectic solvents (NADES) made mainly with abundant primary metabolites are being increasingly applied in green chemistry. The advantages of NADES as green solvents have led to their use in novel green products for the food, cosmetics and pharma
Dan Zhao et al.
Nanomaterials (Basel, Switzerland), 8(8) (2018-08-15)
The recognition of fingerprints has played an extremely important role in criminal investigations, due to its uniqueness. This paper reports on the recognition of sweat latent fingerprints using green-emitting, environment-friendly carbon dots prepared with DL-malic acid and ethylenediamine, and the
G Anneroth et al.
Scandinavian journal of dental research, 88(3), 214-218 (1980-06-01)
Two saliva stimulating drugs, solutions of Hybrin and malic acid were in an in vitro study found to have a heavy demineralizing effect on the root surface of the dental hard tissue. Consequently the drugs cannot be recommended to patients
Hugo Oliveira et al.
PloS one, 9(10), e108376-e108376 (2014-10-08)
Resistance rates are increasing among several problematic Gram-negative pathogens, a fact that has encouraged the development of new antimicrobial agents. This paper characterizes a Salmonella phage endolysin (Lys68) and demonstrates its potential antimicrobial effectiveness when combined with organic acids towards
Cuiyue Liang et al.
Plant physiology, 161(3), 1347-1361 (2013-01-24)
Low pH, aluminum (Al) toxicity, and low phosphorus (P) often coexist and are heterogeneously distributed in acid soils. To date, the underlying mechanisms of crop adaptation to these multiple factors on acid soils remain poorly understood. In this study, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service