Skip to Content
Merck
All Photos(1)

Key Documents

15-2380

Sigma-Aldrich

Isopropylamine

SAJ special grade, ≥99.0%

Synonym(s):

2-Aminopropane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHNH2
CAS Number:
Molecular Weight:
59.11
Beilstein:
605259
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
grade:
SAJ special grade
bp:
33-34 °C (lit.)
vapor pressure:
9.2 psi ( 20 °C)
Pricing and availability is not currently available.

grade

SAJ special grade

vapor density

2.04 (vs air)

vapor pressure

9.2 psi ( 20 °C)

Assay

≥99.0%

autoignition temp.

755 °F

expl. lim.

10.4 %

availability

available only in Japan

refractive index

n20/D 1.374 (lit.)

bp

33-34 °C (lit.)

density

0.688 g/mL at 20 °C (lit.)

SMILES string

CC(C)N

InChI

1S/C3H9N/c1-3(2)4/h3H,4H2,1-2H3

InChI key

JJWLVOIRVHMVIS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 1 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

<-13.0 °F - closed cup

Flash Point(C)

<= -25 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Omar Deeb et al.
Chemical biology & drug design, 71(4), 352-362 (2008-03-04)
The aim of this study is to provide an initial indication regarding the scope and limitations of some state-of-the-art methods in computational chemistry, including semiempirical (AM1) and density functional theory (B3LYP), in the flip regression procedure applied to the inhibition
Karim Engelmark Cassimjee et al.
Chemical communications (Cambridge, England), 46(30), 5569-5571 (2010-05-13)
Enantiopure chiral amines synthesis using omega-transaminases is hindered by an unfavourable equilibrium, but when using isopropylamine as the amine donor the equilibrium can be completely displaced by using a specific dehydrogenase in situ for removal of formed acetone.
Bambang Veriansyah et al.
Journal of hazardous materials, 124(1-3), 119-124 (2005-06-09)
Supercritical water oxidation can effectively destroy a large variety of high-risk wastes resulting from munitions demilitarization and complex industrial chemical. An important design consideration in the development of supercritical water oxidation is the information on the oxidation rate. In this
Katrijn De Klerck et al.
Journal of chromatography. A, 1234, 72-79 (2011-12-07)
In chiral supercritical fluid chromatography (SFC), mobile-phase additives are often used to improve enantioseparations and peak shapes. An acidic or basic additive is chosen, depending on the nature of the compound. This work highlights the simultaneous use of the acidic
Gabriel Vallejos et al.
Bioorganic & medicinal chemistry, 13(14), 4450-4457 (2005-05-24)
The in vitro monoamine oxidase inhibitory (MAOI) activities of 11 heteroarylisopropylamines vis-à-vis MAO-A and MAO-B were described and interpreted in terms of possible interactions with the enzyme active site. Molecular dynamics simulations allowed a comparison between the most active MAO-A

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service