Skip to Content
Merck
All Photos(1)

Documents

15-1480

Sigma-Aldrich

Iron(III) sulfate hydrate

SAJ first grade, 60.0-80.0% anhydrous basis

Synonym(s):

Ferric sulfate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Fe2(SO4)3 · xH2O
CAS Number:
Molecular Weight:
399.88 (anhydrous basis)
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

SAJ first grade

Assay

60.0-80.0% anhydrous basis

form

powder

reaction suitability

reagent type: catalyst
core: iron

availability

available only in Japan

storage temp.

15-25°C

SMILES string

[Fe+3].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/2Fe.3H2O4S/c;;3*1-5(2,3)4/h;;3*(H2,1,2,3,4)/q2*+3;;;/p-6

InChI key

RUTXIHLAWFEWGM-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Y Zhang et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 64(7), 1413-1418 (2011-12-20)
This work aims to investigate removal efficiency of oxidation and coagulation/flocculation processes, to provide an effective method for the treatment of biologically pre-treated leachate. Leachate containing 985 mg L(-1) COD was treated by using three treatment schemes, i.e. oxidation, coagulation/flocculation
M E Odabaş et al.
European journal of paediatric dentistry : official journal of European Academy of Paediatric Dentistry, 13(2), 118-122 (2012-07-06)
The purpose of this study was to evaluate and compare the clinical and radiographic findings of ferric sulphate (FS) and mineral trioxide aggregate (MTA) as vital pulpotomy materials in primary molars. Participants, including 40 boys and 53 girls ranging between
Gal Frenkel et al.
The Journal of clinical pediatric dentistry, 37(2), 137-141 (2012-01-01)
To compare the long-term clinical and radiographic outcomes of pulpotomies in primary molars performed with white or gray Mineral Trioxide Aggregate (MTA) in combination with ferric sulfate (FS), when one package of MTA is used for multiple treatments. Sixty eight
Antonio Scarano et al.
Journal of endodontics, 38(1), 20-23 (2011-12-14)
Calcium sulfate (CaS) is a simple, biocompatible material with a long history of safe use in different fields of medicine. CaS is a rapidly resorbing material that leaves behind a calcium phosphate lattice, which promotes bone regeneration and hemostasis. The
Sudip Bhattacharyya et al.
Free radical research, 46(10), 1296-1307 (2012-07-24)
In the present study we investigated the beneficial role of glycine in iron (FeSO₄) induced oxidative damage in murine hepatocytes. Exposure of hepatocytes to 20 μM FeSO₄ for 3 hours enhanced reactive oxygen species (ROS) generation and induced alteration in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service