Skip to Content
Merck
All Photos(1)

Key Documents

13-2220

Sigma-Aldrich

HEPES

≥99.0%, solid

Synonym(s):

4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H18N2O4S
CAS Number:
Molecular Weight:
238.30
Beilstein:
883043
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:

Product Name

HEPES, ≥99.0%

Assay

≥99.0%

form

solid

availability

available only in Japan

useful pH range

6.8-8.2

pKa (25 °C)

7.5

SMILES string

OCCN1CCN(CC1)CCS(O)(=O)=O

InChI

1S/C8H18N2O4S/c11-7-5-9-1-3-10(4-2-9)6-8-15(12,13)14/h11H,1-8H2,(H,12,13,14)

InChI key

JKMHFZQWWAIEOD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Suitability

for biological purposes

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Dey et al.
Oncogene, 33(33), 4213-4225 (2013-10-01)
Estrogen receptor β (ERβ) is emerging as a critical factor in understanding prostate cancer biology. Although reduced in prostate cancer above Gleason grade 3, ERβ is a potential drug target at the initial stage of the disease. In human prostate
Frédéric Perros et al.
Journal of the American College of Cardiology, 65(7), 668-680 (2015-02-14)
Endothelial cell (EC) dysfunction plays a central role in the pathogenesis of pulmonary arterial hypertension (PAH), promoting vasoconstriction, smooth muscle proliferation, and inflammation. This study sought to test the hypothesis that nebivolol, a β1-antagonist and β2,3-agonist, may improve PAH and
Alexandre Albanese et al.
ACS nano, 8(6), 5515-5526 (2014-05-07)
A nanoparticle's physical and chemical properties at the time of cell contact will determine the ensuing cellular response. Aggregation and the formation of a protein corona in the extracellular environment will alter nanoparticle size, shape, and surface properties, giving it
Suganthi Chittaranjan et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 20(12), 3159-3173 (2014-04-12)
Triple-negative breast cancers (TNBC) are defined by a lack of expression of estrogen receptor (ER), progesterone receptor (PR), and human epidermal growth factor receptor 2 (ERBB2/HER2). Although initially responsive to chemotherapy, most recurrent TNBCs develop resistance, resulting in disease progression.
Bérangère Pinan-Lucarré et al.
Nature, 511(7510), 466-470 (2014-06-05)
Because most neurons receive thousands of synaptic inputs, the neuronal membrane is a mosaic of specialized microdomains where neurotransmitter receptors cluster in register with the corresponding presynaptic neurotransmitter release sites. In many cases the coordinated differentiation of presynaptic and postsynaptic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service