13-1100
β-Thujaplicin
SAJ special grade, ≥99.0%
Synonym(s):
2-Hydroxy-4-isopropyl-2,4,6-cycloheptatrien-1-one, Hinokitiol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H12O2
CAS Number:
Molecular Weight:
164.20
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Recommended Products
grade
SAJ special grade
Assay
≥99.0%
availability
available only in Japan
bp
140 °C/10 mmHg (lit.)
mp
50-52 °C (lit.)
SMILES string
CC(C)C1=CC=CC(=O)C(O)=C1
InChI
1S/C10H12O2/c1-7(2)8-4-3-5-9(11)10(12)6-8/h3-7H,1-2H3,(H,11,12)
InChI key
FUWUEFKEXZQKKA-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Yu-Jing Zhu et al.
Journal of enzyme inhibition and medicinal chemistry, 25(6), 798-803 (2010-06-29)
The inhibitory effects of hinokitiol, a constituent of the woody oils isolated from Cupressaceae heartwood, on mushroom tyrosinase and melanin formation in B16 melanoma cells as well as its antimicrobial activity were investigated. Our results showed that hinokitiol could strongly
S L Hwang et al.
Journal of microencapsulation, 25(5), 351-356 (2008-05-10)
Nanocapsules containing hinokitiol (HKL) were prepared by an emulsion-diffusion method. In an emulsification step in preparing nanocapsules, cetyltrimethylamonium chloride (CTAC) was employed as a cationic emulsifier, Poly(epsilon-caprolactone) (PCL) was use as a wall material and HKL dissolved in octylsalicylate (OS)
Tsair-Bor Yen et al.
Bioresource technology, 99(11), 4871-4877 (2007-11-06)
The ethanolic extract of Calocedrus macrolepis var. formosana heartwood was screened for antifungal compounds by agar dilution assay and liquid chromatography. Two compounds, beta-thujaplicin and gamma-thujaplicin, responsible for the antifungal property of C. macrolepis var. formosana heartwood were isolated by
Se Eun Byeon et al.
Planta medica, 74(8), 828-833 (2008-06-10)
In this study, we examined the modulatory effect of hinokitiol (HK) on the production of tumor necrosis factor (TNF)-alpha, a critical factor involved in skin inflammation and hair follicle apoptosis. HK effectively suppressed TNF-alpha production in lipopolysaccharide (LPS)-activated, macrophage-like (RAW264.7)
Ransika De Alwis et al.
Journal of plant physiology, 166(7), 720-728 (2008-11-26)
Elicitor treatment initiates defense responses in cultured Cupressus lusitanica cells. In order to investigate the defense mechanism with a yeast extract elicitor, we carried out solid-phase microextraction coupled with gas chromatography for monoterpene analysis. Ten hydrocarbon monoterpenes, including high amounts
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service