Skip to Content
Merck
All Photos(1)

Key Documents

13-0940

Sigma-Aldrich

1-Hexanol

SAJ special grade, ≥99.0%

Synonym(s):

Hexyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5OH
CAS Number:
Molecular Weight:
102.17
Beilstein:
969167
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
grade:
SAJ special grade
Assay:
≥99.0%
bp:
156-157 °C (lit.)
vapor pressure:
1 mmHg ( 25.6 °C)

grade

SAJ special grade

vapor density

4.5 (vs air)

vapor pressure

1 mmHg ( 25.6 °C)

Assay

≥99.0%

form

liquid

autoignition temp.

559 °F

expl. lim.

0.34-6.3 % (lit.)

availability

available only in Japan

refractive index

n20/D 1.418 (lit.)

bp

156-157 °C (lit.)

mp

−52 °C (lit.)

density

0.814 g/mL at 25 °C (lit.)

SMILES string

CCCCCCO

InChI

1S/C6H14O/c1-2-3-4-5-6-7/h7H,2-6H2,1H3

InChI key

ZSIAUFGUXNUGDI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

140.0 °F - closed cup

Flash Point(C)

60 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mir Ali Farajzadeh et al.
Analytica chimica acta, 713, 70-78 (2011-12-28)
In the present work a new, simple, rapid and environmentally friendly dispersive liquid-liquid microextraction (DLLME) method has been developed for extraction/preconcentration of some triazole pesticides in aqueous samples and in grape juice. The extract was analyzed with gas chromatography-flame ionization
Zhisong Lu et al.
Nanotechnology, 22(15), 155604-155604 (2011-03-11)
With advances of quantum dots (QDs) in bioimaging applications, various materials have been used to coat QDs to reduce their nanotoxicity; however, the coating could introduce new toxic sources and quench the fluorescence in bioimaging applications. In this work, ZrO₂
Yasumasa Dekishima et al.
Journal of the American Chemical Society, 133(30), 11399-11401 (2011-06-29)
An Escherichia coli strain was engineered to synthesize 1-hexanol from glucose by extending the coenzyme A (CoA)-dependent 1-butanol synthesis reaction sequence catalyzed by exogenous enzymes. The C4-acyl-CoA intermediates were first synthesized via acetyl-CoA acetyltransferase (AtoB), 3-hydroxybutyryl-CoA dehydrogenase (Hbd), crotonase (Crt)
Abdolhamid Firooz et al.
Journal of colloid and interface science, 370(1), 183-191 (2012-01-27)
The effects of carbon chain length and temperature were investigated on adsorption kinetics and surface tension of a group of slightly volatile, short carbon chain molecules: 1-octanol, 1-hexanol, and 1-butanol. Experiments were performed in a closed chamber where simultaneous adsorption
Carolina Aubery et al.
Langmuir : the ACS journal of surfaces and colloids, 29(6), 1779-1789 (2013-01-12)
Phase behavior, dynamics, and structure of W/O microemulsions of the system aqueous solution/Synperonic 13_6.5/1-hexanol/isooctane were studied, with the goal of determining their effect on Mn-Zn ferrite nanoparticle formation, kinetics and characteristics. Microemulsion structure and dynamics were studied systematically by conductivity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service