Skip to Content
Merck
All Photos(1)

Documents

12-1521

Sigma-Aldrich

Gram-Hucker′s stain solution (I)

Synonym(s):

Gentian Violet solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116130
PubChem Substance ID:

availability

available only in Japan

SMILES string

[Cl-].CN(C)c1ccc(cc1)\C(c2ccc(cc2)N(C)C)=C3/C=C\C(C=C3)=[N+](/C)C

InChI

1S/C25H30N3.ClH/c1-26(2)22-13-7-19(8-14-22)25(20-9-15-23(16-10-20)27(3)4)21-11-17-24(18-12-21)28(5)6;/h7-18H,1-6H3;1H/q+1;/p-1

InChI key

ZXJXZNDDNMQXFV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Components

Composition : Crystal violet in water

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Henry M Wu et al.
BMC infectious diseases, 13, 26-26 (2013-01-24)
Although cerebrospinal fluid (CSF) culture is the diagnostic reference standard for bacterial meningitis, its sensitivity is limited, particularly when antibiotics were previously administered. CSF Gram staining and real-time PCR are theoretically less affected by antibiotics; however, it is difficult to
Antimicrobial stewardship lessons: know when to say no to vancomycin.
Rodrigo Pires dos Santos et al.
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America, 56(4), 616-617 (2012-10-24)
Lise Crémet et al.
Diagnostic microbiology and infectious disease, 75(3), 252-255 (2013-01-15)
Biofilm formation seems to be a key factor in many bacterial infections, particularly those involving prosthetic implants or urinary catheters, where Escherichia coli is frequently involved. We have determined the ability to form biofilm in vitro of 34 E. coli
Leilani Ireland Way et al.
Journal of veterinary emergency and critical care (San Antonio, Tex. : 2001), 23(1), 23-28 (2013-01-16)
To determine the utility of performing urine Gram stain for detection of bacteriuria compared to routine urine sediment examination and bacterial aerobic urine culture. Prospective, observational study. University teaching hospital. Urine samples acquired via cystocentesis through convenience sampling from 103
Gram stain for Vibrio species.
Chia-Chang Kuo et al.
The American journal of emergency medicine, 31(5), 877-878 (2013-03-19)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service