Skip to Content
Merck
All Photos(1)

Key Documents

09-1689

Sigma-Aldrich

2-Ethoxyethanol

SAJ first grade, ≥97.0%

Synonym(s):

Ethyl glycol, Ethylene glycol monoethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCH2CH2OH
CAS Number:
Molecular Weight:
90.12
Beilstein:
1098271
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:

grade

SAJ first grade

vapor density

3.1 (vs air)

vapor pressure

3.8 mmHg ( 20 °C)

Assay

≥97.0%

form

liquid

autoignition temp.

460 °F

expl. lim.

14 %

availability

available only in Japan

refractive index

n20/D 1.407 (lit.)

bp

135 °C (lit.)

mp

−90 °C (lit.)

density

0.93 g/mL at 25 °C (lit.)

SMILES string

CCOCCO

InChI

1S/C4H10O2/c1-2-6-4-3-5/h5H,2-4H2,1H3

InChI key

ZNQVEEAIQZEUHB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3 - Repr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

111.2 °F

Flash Point(C)

44 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rui-Sheng Wang et al.
Industrial health, 45(4), 574-578 (2007-09-20)
Ethylene glycol monoethyl ether (EGEE) can cause damage to testes and sperm, and its metabolites are believed to play an important role in its toxicity. Aldehyde dehydrogenase 2 (ALDH2) is involved in the metabolism of this chemical. To investigate whether
Isaac A Adedara et al.
Human & experimental toxicology, 29(10), 801-812 (2010-02-23)
The effects of ethylene glycol monoethyl ether (EGEE) on the antioxidant systems of the testes and epididymal spermatozoa were investigated in rats at dose levels of 0, 100, 200 and 400 mg kg(-1) body weight (bw) administered orally by gavage
I A Adedara et al.
Human & experimental toxicology, 31(5), 506-517 (2011-10-27)
The present study investigated the protective effect of kolaviron, a biflavonoid from the seed of Garcinia kola, on ethylene glycol monoethyl ether (EGEE)-induced reproductive toxicity in male rats. The protective effect of kolaviron was validated using vitamin E, a standard
Randa A El-Zein et al.
Archives of environmental health, 57(4), 371-376 (2003-01-18)
Glycol ethers are known reproductive and developmental toxins in laboratory animals, but little is known about their genotoxic effects in humans. In the current article, the authors tested the hypothesis that human in utero exposure to ethylene glycol monomethyl ether
David J Lockley et al.
Toxicology and applied pharmacology, 180(2), 74-82 (2002-04-24)
Percutaneous absorption and cutaneous metabolism of 2-ethoxyethanol were assessed in vivo and with an in vitro flow-through diffusion system. Topical application of undiluted (14)C-ethoxyethanol to occluded rat skin in vivo resulted in 25% of the dose being absorbed after 24

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service