Skip to Content
Merck
All Photos(1)

Documents

05-6500

Sigma-Aldrich

p-Cresol

JIS special grade, ≥99.0%

Synonym(s):

4-Methylphenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4OH
CAS Number:
Molecular Weight:
108.14
Beilstein:
1305151
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

grade

JIS special grade

vapor density

3.72 (vs air)

vapor pressure

1 mmHg ( 20 °C)

Assay

≥99.0%

form

crystalline

autoignition temp.

1038 °F

expl. lim.

1 %
1.1 %, 150 °F

availability

available only in Japan

bp

202 °C (lit.)

mp

32-34 °C (lit.)

density

1.034 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(O)cc1

InChI

1S/C7H8O/c1-6-2-4-7(8)5-3-6/h2-5,8H,1H3

InChI key

IWDCLRJOBJJRNH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

186.8 °F - closed cup

Flash Point(C)

86 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fangxin Hu et al.
Analytica chimica acta, 724, 40-46 (2012-04-10)
In this paper, the reduced graphene oxide and multiwall carbon nanotubes hybrid materials (RGO-MWNTs) were prepared and a strategy for detecting environmental contaminations was proposed on the basis of RGO-MWNTs modified electrode. The hybrid materials were characterized by the scanning
K Akasaka et al.
Journal of chromatography, 617(2), 205-211 (1993-08-11)
Cholesterol ester (ChE) and triacylglycerol (TG) hydroperoxides in human plasma were determined by high-performance liquid chromatography with postcolumn detection with diphenyl-1-pyrenylphosphine. Human plasma was extracted once with n-hexane. 2,6-Di-tert.-butyl-4-methylphenol and N-stearylcinnamide were added to human plasma before extraction as an
P Bühlmann et al.
Analytical chemistry, 73(14), 3199-3205 (2001-07-31)
Ionophore-free ion exchanger electrodes were found to exhibit quite a high selectivity for the creatininium ion; however, measurements in diluted urine samples revealed large emf drifts. Potentiometric, chromatographic, NMR, and mass spectrometric evidence did not reveal any major cationic interfering
Bert Bammens et al.
Kidney international, 64(6), 2196-2203 (2003-11-25)
Protein malnutrition is a common finding in chronic renal failure (CRF) and is associated with poor outcome. We hypothesized that besides inadequate dietary protein intake and alterations in metabolism, deficient protein assimilation (digestion and absorption) might contribute to the pathogenesis
Isabelle E J A François et al.
The British journal of nutrition, 108(12), 2229-2242 (2012-03-01)
Wheat bran extract (WBE) is a food-grade soluble fibre preparation that is highly enriched in arabinoxylan oligosaccharides. In this placebo-controlled cross-over human intervention trial, tolerance and effects on colonic protein and carbohydrate fermentation were studied. After a 1-week run-in period

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service