Skip to Content
Merck
All Photos(1)

Key Documents

05-6470

Sigma-Aldrich

o-Cresol

SAJ first grade, ≥97.0%

Synonym(s):

2-Methylphenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4OH
CAS Number:
Molecular Weight:
108.14
Beilstein:
506917
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
Pricing and availability is not currently available.

grade

SAJ first grade

vapor density

3.72 (vs air)

vapor pressure

0.3 mmHg ( 20 °C)

Assay

≥97.0%

form

powder

autoignition temp.

1110 °F

expl. lim.

1.47 %, 148 °F

availability

available only in Japan

pH

4.5 (25 g/L)

bp

191 °C (lit.)

mp

29-31 °C (lit.)

SMILES string

Cc1ccccc1O

InChI

1S/C7H8O/c1-6-4-2-3-5-7(6)8/h2-5,8H,1H3

InChI key

QWVGKYWNOKOFNN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hua-Wei Chen et al.
Chemosphere, 69(2), 184-190 (2007-06-05)
This study investigated the effect of controlled periodic illumination on the photonic efficiency and temporal decomposition behavior of o-cresol by UV/TiO2 process in a slurry reactor using light emitting diodes as the light source. Based on the same UV light
Wagdy K B Khalil et al.
Arhiv za higijenu rada i toksikologiju, 62(2), 121-129 (2011-06-28)
Cresols are monomethyl derivatives of phenol frequently used as solvents and intermediates in the production of disinfectants, fragrances, pesticides, dyes, and explosives, which is probably why they are widely distributed in the environment. General population may be exposed to cresols
G Koppen et al.
Journal of applied toxicology : JAT, 27(3), 238-246 (2007-01-18)
The present paper deals with the evaluation of a battery of genotoxicity biomarkers in healthy Flemish adolescents and their relation with common pollutants occurring in their life environment. DNA damage as reflected by the comet assay appeared to be most
María Eugenia Suarez-Ojeda et al.
Chemosphere, 66(11), 2096-2105 (2006-11-11)
This study examines the feasibility of coupling a Catalytic Wet Air Oxidation (CWAO), with activated carbon (AC) as catalyst, and an aerobic biological treatment to treat a high-strength o-cresol wastewater. Two goals are pursued: (a) To determine the effect of
Johanna M Dela Cruz et al.
Journal of mass spectrometry : JMS, 42(2), 178-186 (2006-12-19)
The influence shaped femtosecond laser pulses have on molecular photofragmentation and ionization, coupled with the intrinsic sensitivity of mass spectrometry, results in a powerful tool for fast, accurate, reproducible and quantitative isomeric identification. Complex phase functions are introduced to enhance

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service