Skip to Content
Merck
All Photos(1)

Key Documents

05-5895

Sigma-Aldrich

Copper(II) chloride

SAJ first grade, ≥98.0%

Synonym(s):

Cupric chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CuCl2
CAS Number:
Molecular Weight:
134.45
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

SAJ first grade

Assay

≥98.0%

form

solid

reaction suitability

reagent type: catalyst
core: copper

availability

available only in Japan

mp

620 °C (lit.)

density

3.386 g/mL at 25 °C (lit.)

storage temp.

15-25°C

SMILES string

Cl[Cu]Cl

InChI

1S/2ClH.Cu/h2*1H;/q;;+2/p-2

InChI key

ORTQZVOHEJQUHG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kenji Funaki et al.
Organic letters, 14(24), 6186-6189 (2012-12-12)
Direct arylation of thiophenes and benzothiophenes with aryltrimethylsilanes was effectively catalyzed by PdCl(2)(MeCN)(2) in the presence of CuCl(2) as an oxidant. The reaction preferentially occurred at the β-position of both thiophenes and benzothiophenes.
Mónica João Barros Amorim et al.
Environmental pollution (Barking, Essex : 1987), 164, 164-168 (2012-03-01)
Environmental effects of copper nanoparticles are little studied in terrestrial ecosystems. In the present article, the toxicity of copper nanoparticles (Cu-NP) on the enchytraeid Enchytraeus albidus is compared to the toxicity of a copper-salt (CuCl(2)). The effect parameters studied were
Iron(II)-catalyzed trifluoromethylation of potassium vinyltrifluoroborates.
Andrew T Parsons et al.
Angewandte Chemie (International ed. in English), 51(12), 2947-2950 (2012-02-14)
Francesca Cima et al.
Chemosphere, 89(1), 19-29 (2012-05-01)
After the widespread ban of TBT, due to its severe impact on coastal biocoenoses, mainly related to its immunosuppressive effects on both invertebrates and vertebrates, alternative biocides such as Cu(I) salts and the triazine Irgarol 1051, the latter previously used
Pieter Spincemaille et al.
Toxicology and applied pharmacology, 280(2), 345-351 (2014-08-20)
Wilson disease (WD) is caused by accumulation of excess copper (Cu) due to a mutation in the gene encoding the liver Cu transporter ATP7B, and is characterized by acute liver failure or cirrhosis and neuronal cell death. We investigated the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service