Skip to Content
Merck
All Photos(1)

Documents

05-3050

Sigma-Aldrich

Chlorobenzene

SAJ first grade, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5Cl
CAS Number:
Molecular Weight:
112.56
Beilstein:
605632
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

grade

SAJ first grade

vapor density

3.86 (vs air)

vapor pressure

11.8 mmHg ( 25 °C)

Assay

≥99.0%

form

liquid

autoignition temp.

1178 °F

expl. lim.

7.1 %

availability

available only in Japan

refractive index

n20/D 1.524 (lit.)

pH

7

bp

132 °C (lit.)

mp

−45 °C (lit.)

density

1.106 g/mL at 25 °C (lit.)

SMILES string

Clc1ccccc1

InChI

1S/C6H5Cl/c7-6-4-2-1-3-5-6/h1-5H

InChI key

MVPPADPHJFYWMZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

80.6 °F

Flash Point(C)

27 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yubo Wang et al.
Nature communications, 11(1), 6089-6089 (2020-12-02)
Nano-sized metal particles are attracting much interest in industrial and biomedical applications due to the recent progress and development of nanotechnology, and the surface-modifications by appropriate polymers are key techniques to stably express their characteristics. Herein, we applied cyclic poly(ethylene
Wei-Chao Chen et al.
ACS applied materials & interfaces, 7(8), 4616-4622 (2015-02-11)
Controlled dispersion of single-walled carbon nanotubes (SWCNTs) in common solvents is a challenging issue, especially for the rising need of low cost flexible transparent conducting films (TCFs). Utilizing conductive polymer as surfactant to facilitate SWCNTs solubility is the most successful
Mi Yan et al.
Waste management (New York, N.Y.), 32(7), 1453-1458 (2012-04-06)
Nitrogen containing compounds such as ammonia, urea and amines can effectively inhibit the formation of polychlorinated dibenzo-p-dioxins and dibenzofurans (PCDD/Fs). Sewage sludge accumulates both sulfur and nitrogen during wastewater treatment so it could be used to reduce PCDD/Fs formation. Indeed
M Govindarajan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 88, 37-48 (2011-12-27)
In this work, the vibrational spectral analysis was carried out by using FT-Raman and FT-IR spectroscopy in the range 100-4000 cm(-1) and 400-4000 cm(-1) respectively, for the title molecule. The molecular structure, fundamental vibrational frequencies and intensity of the vibrational
Kerry N McPhedran et al.
Water research, 47(7), 2222-2230 (2013-03-12)
The sorption behaviour of hydrophobic organic compounds (HOCs) 1,2,4,5-tetrachlor-obenzene (TeCB), pentachlorobenzene (PeCB) and hexachlorobenzene (HCB) to Aldrich humic acid (AHA) and municipal wastewater treatment plant (MWTP) influent colloidal organic carbon (COC) was investigated using the gas-stripping technique. Gas stripping assumptions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service