Skip to Content
Merck
All Photos(1)

Key Documents

05-2620

Sigma-Aldrich

Charcoal

from bovine (calf) bone, CP

Synonym(s):

Activated charcoal, Charcoal activated

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
EC Number:
MDL number:
UNSPSC Code:
12000000

biological source

bovine (calf) bone

grade

CP

vapor pressure

<0.1 mmHg ( 20 °C)

form

powder

autoignition temp.

842 °F

availability

available only in Japan

resistivity

1375 μΩ-cm, 20°C (graphite)

mp

3550 °C (lit.)

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher D Cappa et al.
Science (New York, N.Y.), 339(6118), 393-393 (2013-09-04)
Jacobson argues that our statement that "many climate models may overestimate warming by BC" has not been demonstrated. Jacobson challenges our results on the basis that we have misinterpreted some model results, omitted optical focusing under high relative humidity conditions
I N Kurchenko et al.
Mikrobiolohichnyi zhurnal (Kiev, Ukraine : 1993), 75(3), 12-23 (2013-07-23)
The ability of soil and endophytic strains of P. funiculosum to accumulate biomass under cultivation conditions in media containing carbon sources from mono- to polysaccharides has been studied. It has been shown that the most favorable carbon sources for the
Fei Tian et al.
Journal of nanoscience and nanotechnology, 13(6), 4468-4471 (2013-07-19)
Hydrophilic carbon nanoonions coated with carbon nanobubbles were synthesized by long-pulse-width laser ablating of carbon black particles suspended in water. The high resolution TEM (HTREM), selected area electron diffraction (SAED) and Raman spectrometer results revealed that the crystallinity and the
Melina Nisenbaum et al.
Journal of environmental sciences (China), 25(3), 613-625 (2013-08-08)
We report on the biodegradation of pure hydrocarbons and chemotaxis towards these compounds by an isolated chlorophenol degrader, Pseudomonas strain H. The biochemical and phylogenetic analysis of the 16S rDNA sequence identified Pseudomonas strain H as having 99.56% similarity with
T Southorn et al.
Journal of perioperative practice, 23(6), 144-146 (2013-08-06)
Currently, there are very few provisions for recycling in theatres. We measured the weight of clinical waste for several orthopaedic operations. This waste was then examined and sorted into domestic waste and clinical waste. With staff education it is possible

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service