Skip to Content
Merck
All Photos(1)

Key Documents

05-1080

Sigma-Aldrich

Calcium sulfate hemihydrate

CP, calcined

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaSO4 · 0.5H2O
CAS Number:
Molecular Weight:
145.15
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

CP

form

calcined

availability

available only in Japan

storage temp.

15-25°C

SMILES string

O.[Ca++].[Ca++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/2Ca.2H2O4S.H2O/c;;2*1-5(2,3)4;/h;;2*(H2,1,2,3,4);1H2/q2*+2;;;/p-4

InChI key

ZOMBKNNSYQHRCA-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Massimo Peruffo et al.
Physical chemistry chemical physics : PCCP, 15(6), 1956-1965 (2012-12-22)
Dissolution processes at single crystal surfaces often involve the initial formation and expansion of localized, characteristic (faceted) etch-pits at defects, in an otherwise comparatively unreactive surface. Using natural gypsum single crystal as an example, a simple but powerful morphological analysis
Rawaa Ammar et al.
Environmental science and pollution research international, 20(12), 9014-9025 (2013-06-15)
One of the most important sources of solid waste in the Mediterranean Basin ecosystem originated from the phosphate fertilizer industries, which discharge phosphogypsum (PG) directly into aquatic environments or are stacked on stockpiles. The present study investigates metal release from
C M Belfiore et al.
Environmental science and pollution research international, 20(12), 8848-8859 (2013-06-08)
Black crusts are recognized to have been, up to now, one of the major deterioration forms affecting the built heritage in urban areas. Their formation is demonstrated to occur mainly on carbonate building materials, whose interaction with an SO2-loaded atmosphere
So-Yeun Kim et al.
The International journal of prosthodontics, 26(2), 161-163 (2013-03-12)
The purpose of this investigation was to measure the accuracy of digital impressions (DIs) compared to conventional impressions (CIs). Using the iTero system, a master cast was scanned to produce stereolithography dies. As a control group, silicone impressions were taken
Eun Young Rha et al.
Plastic and reconstructive surgery, 133(1), 14-20 (2014-01-01)
The authors have developed a simple and intuitive method for measuring breast volume based on three-dimensional simulated images of magnetic resonance imaging scans to accurately estimate breast volume before breast reconstruction. The authors performed a retrospective review of the medical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service