Skip to Content
Merck
All Photos(1)

Documents

05-0760

Sigma-Aldrich

Calcium hydroxide

CP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ca(OH)2
CAS Number:
Molecular Weight:
74.09
EC Number:
MDL number:
UNSPSC Code:
12352305
PubChem Substance ID:

grade

CP

form

powder

availability

available only in Japan

pH

12.4-12.6 (20 °C)

density

2.24 g/mL at 25 °C (lit.)

SMILES string

O[Ca]O

InChI

1S/Ca.2H2O/h;2*1H2/q+2;;/p-2

InChI key

AXCZMVOFGPJBDE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nikte Gómez-Ortíz et al.
ACS applied materials & interfaces, 5(5), 1556-1565 (2013-01-26)
The presence and deteriorating action of microbial biofilms on historic stone buildings have received considerable attention in the past few years. Among microorganisms, fungi are one of the most damaging groups. In the present work, antimicrobial surfaces were prepared using
Lisbet Mailin López González et al.
Bioresource technology, 131, 250-257 (2013-01-29)
Different pre-treatment severities by thermo-alkaline conditions (100°C, Ca(OH)2) on press mud were evaluated for different pre-treatment time and lime loading. COD solubilization and the methane yield enhancement were assessed. The biochemical methane potential was determined in batch assays under mesophilic
P Sangwan et al.
International endodontic journal, 46(1), 3-19 (2012-08-15)
Calcium hydroxide has been used extensively in dentistry for a century. Despite its widespread use as a pulp-capping agent, its mechanisms of action still remain ambiguous. Understanding its modes of action will lead to a broader understanding of the mechanisms
Z Mohammadi et al.
International endodontic journal, 44(8), 697-730 (2011-05-04)
Calcium hydroxide has been included within several materials and antimicrobial formulations that are used in a number of treatment modalities in endodontics. These include, inter-appointment intracanal medicaments, pulp-capping agents and root canal sealers. Calcium hydroxide formulations are also used during
J F Siqueira et al.
International endodontic journal, 32(5), 361-369 (1999-11-07)
Literature review The primary function of calcium hydroxide as a routine intracanal medicament is to provide antimicrobial activity. However, the mechanisms of antimicrobial activity of calcium hydroxide are not well known. Physicochemical properties of this substance may limit its effectiveness

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service