Skip to Content
Merck
All Photos(1)

Key Documents

05-0490

Sigma-Aldrich

Calcium carbonate

SAJ first grade, ≥98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaCO3
CAS Number:
Molecular Weight:
100.09
Beilstein:
8008338
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

SAJ first grade

Assay

≥98.0%

form

powder

availability

available only in Japan

pH

8

density

2.93 g/mL at 25 °C (lit.)

SMILES string

[Ca++].[O-]C([O-])=O

InChI

1S/CH2O3.Ca/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2

InChI key

VTYYLEPIZMXCLO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sophie A Jamal et al.
Lancet (London, England), 382(9900), 1268-1277 (2013-07-23)
Phosphate binders (calcium-based and calcium-free) are recommended to lower serum phosphate and prevent hyperphosphataemia in patients with chronic kidney disease, but their effects on mortality and cardiovascular outcomes are unknown. We aimed to update our meta-analysis on the effect of
Ling Li et al.
Nature materials, 13(5), 501-507 (2014-04-01)
Hierarchical composite materials design in biological exoskeletons achieves penetration resistance through a variety of energy-dissipating mechanisms while simultaneously balancing the need for damage localization to avoid compromising the mechanical integrity of the entire structure and to maintain multi-hit capability. Here
Polysaccharides and proteoglycans in calcium carbonate-based biomineralization.
José L Arias et al.
Chemical reviews, 108(11), 4475-4482 (2008-07-26)
Claudia Gagnon et al.
PloS one, 9(10), e109607-e109607 (2014-10-10)
To examine whether combined vitamin D and calcium supplementation improves insulin sensitivity, insulin secretion, β-cell function, inflammation and metabolic markers. 6-month randomized, placebo-controlled trial. Ninety-five adults with serum 25-hydroxyvitamin D [25(OH)D] ≤55 nmol/L at risk of type 2 diabetes (with
Fei Wang et al.
Environmental science & technology, 49(9), 5672-5680 (2015-04-09)
The mineralization of perfluorinated alkyl substances (PFASs) by calcium compounds during the waste thermal treatment was systemically studied. Different calcium compounds showed different mineralization efficiencies of PFASs during the thermal process, owing to the different reaction mechanisms. Calcium hydroxide was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service