Skip to Content
Merck
All Photos(1)

Key Documents

03-5000

Sigma-Aldrich

Butyl 4-hydroxybenzoate

SAJ first grade, ≥99.0%

Synonym(s):

p-Hydroxybenzoic acid n-butyl ester, Butyl paraben

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CO2(CH2)3CH3
CAS Number:
Molecular Weight:
194.23
Beilstein:
1103741
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

grade

SAJ first grade

Assay

≥99.0%

availability

available only in Japan

bp

156-157 °C/3.5 mmHg (lit.)

mp

67-70 °C (lit.)

SMILES string

CCCCOC(=O)c1ccc(O)cc1

InChI

1S/C11H14O3/c1-2-3-8-14-11(13)9-4-6-10(12)7-5-9/h4-7,12H,2-3,8H2,1H3

InChI key

QFOHBWFCKVYLES-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guoyuan Wen et al.
The Journal of general virology, 96(Pt 6), 1219-1228 (2015-01-30)
Thermostable Newcastle disease virus (NDV) vaccines have been used widely to control Newcastle disease for village poultry flocks, due to their independence of cold chains for delivery and storage. To explore the potential use of thermostable NDV as a vaccine
Jordan Shaw et al.
Reproductive toxicology (Elmsford, N.Y.), 28(1), 26-31 (2009-06-06)
Parabens, a class of preservatives routinely added to cosmetics, pharmaceuticals, and foods, have estrogenic properties. Given that intrauterine implantation of fertilized ova in inseminated females can be disrupted by minute levels of exogenous estrogens, we assessed the impact of parabens
Caroline Stremnitzer et al.
The Journal of investigative dermatology, 135(7), 1790-1800 (2015-02-24)
Papain is commonly used in food, pharmaceutical, textile, and cosmetic industries and is known to induce occupational allergic asthma. We have previously shown that the papain-like cysteine protease Dermatophagoides pteronyssinus 1 from house dust mite exhibits percutaneous sensitization potential. We
Emeran A Mayer et al.
The Journal of clinical investigation, 125(3), 926-938 (2015-02-18)
Tremendous progress has been made in characterizing the bidirectional interactions between the central nervous system, the enteric nervous system, and the gastrointestinal tract. A series of provocative preclinical studies have suggested a prominent role for the gut microbiota in these
Anna Viridén et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 77(1), 99-110 (2010-11-18)
This study investigates the effect of the chemical heterogeneity of hydroxypropyl methylcellulose (HPMC) on the release of model drug substances from hydrophilic matrix tablets. The hypothesis was that the release of drug substances could be influenced by possible interactions with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service