Skip to Content
Merck
All Photos(1)

Documents

03-0160

Sigma-Aldrich

Barium carbonate

SAJ special grade, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BaCO3
CAS Number:
Molecular Weight:
197.34
Beilstein:
7045119
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

SAJ special grade

Assay

≥99.0%

form

powder

availability

available only in Japan

pH

6.8 (37 °C, 3.67 g/L)

SMILES string

[Ba++].[O-]C([O-])=O

InChI

1S/CH2O3.Ba/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2

InChI key

AYJRCSIUFZENHW-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kyeong Eun Song et al.
Frontiers in chemistry, 8, 628813-628813 (2021-03-19)
In SmBa1-xCaxCo2O5+d (x = 0.01, 0.03, 0.1, and 0.2, SBCCO) oxide systems calcined at 1100°C for 8 h, the XRD patterns of the SBCCO single phase were maintained in the cases of SmBa0.97Ca0.03Co2O5+d (SBCCO-0.97) and SmBa0.99Ca0.01Co2O5+d (SBCCO-0.99) compositions. In SmBa0.8Ca0.2Co2O5+d (SBCCO-0.8)
D Ekeberg et al.
Carbohydrate research, 335(2), 141-146 (2001-09-25)
A simple, but low-yielding method for the synthesis of 3-hexuloses has been elaborated. Oxidation of 1,2:5,6-di-O-isopropylidenehexitols with bromine in the presence of barium carbonate, followed by mild-acid hydrolysis of the oxidation products gave the free hexuloses. Oxidation occurred at only
Kevin V Brix et al.
Ecotoxicology and environmental safety, 73(7), 1646-1652 (2010-07-14)
A series of Toxicity Identification Evaluations (TIEs) to identify the cause(s) of observed toxicity to Ceriodaphnia dubia have been conducted on a hard rock mining effluent. Characteristic of hard rock mining discharges, the effluent has elevated (∼3000 mg l(-1)) total
Jian-Hua Zhu et al.
Chemical communications (Cambridge, England), (9)(9), 1106-1108 (2009-02-20)
A facile biomimetic method is reported for the synthesis of novel BaCO(3) nanofibres with double-stranded and cylindrical helical morphologies via a phosphonated block co-polymer-controlled mineralization process.
Ashutosh Dash et al.
Journal of hazardous materials, 168(2-3), 1126-1133 (2009-04-03)
An attempt to recover and purify 4.66 TBq(126Ci) of (137)Cs as solution from a 20-year-old doubly encapsulated sealed source containing (137)CsCl was made successfully. The primary capsule was first cut open to retrieve the secondary capsule. The top end of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service