Skip to Content
Merck
All Photos(1)

Key Documents

01-6730

Sigma-Aldrich

L-Ascorbic acid

JIS special grade, ≥99.0%

Synonym(s):

L-Threoascorbic acid, Antiscorbutic factor, Vitamin C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O6
CAS Number:
Molecular Weight:
176.12
Beilstein:
84272
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
Pricing and availability is not currently available.

grade

JIS special grade

Assay

≥99.0%

form

solid

availability

available only in Japan

mp

190-194 °C (dec.)

solubility

water: soluble 176 g/L at 20 °C

SMILES string

OC([C@]([C@@H](O)CO)([H])O1)=C(O)C1=O

InChI

1S/C6H8O6/c7-1-2(8)5-3(9)4(10)6(11)12-5/h2,5,7-10H,1H2/t2-,5+/m0/s1

InChI key

CIWBSHSKHKDKBQ-JLAZNSOCSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kathryn Blaschke et al.
Nature, 500(7461), 222-226 (2013-07-03)
DNA methylation is a heritable epigenetic modification involved in gene silencing, imprinting, and the suppression of retrotransposons. Global DNA demethylation occurs in the early embryo and the germ line, and may be mediated by Tet (ten eleven translocation) enzymes, which convert 5-methylcytosine
Noriko Umegaki-Arao et al.
Science translational medicine, 6(264), 264ra164-264ra164 (2014-11-28)
Revertant mosaicism is a naturally occurring phenomenon involving spontaneous correction of a pathogenic gene mutation in a somatic cell. It has been observed in several genetic diseases, including epidermolysis bullosa (EB), a group of inherited skin disorders characterized by blistering
Stefanie B Murer et al.
The Journal of nutrition, 144(2), 193-201 (2013-12-20)
Oxidative stress and low-grade systemic inflammation may contribute to the pathogenesis of obesity-induced comorbidities, including nonalcoholic fatty liver disease. Increasing intake of dietary antioxidants might be beneficial, but there are few data in obese children. To examine the effect of
Marc P Bradshaw et al.
Critical reviews in food science and nutrition, 51(6), 479-498 (2011-09-21)
Extensive reviews of research are available on the use of ascorbic acid, and its consequent degradation pathways, in physiological conditions or food matrices. However, very little information can be found for wine-related systems. This review highlights the relevant chemistry and
Zi Yin et al.
Biomaterials, 44, 173-185 (2015-01-27)
Physical topographic cues from various substrata have been shown to exert profound effects on the growth and differentiation of stem cells due to their niche-mimicking features. However, the biological function of different topographic materials utilized as bio-scaffolds in vivo have

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service