Skip to Content
Merck
All Photos(1)

Key Documents

01-6635

Sigma-Aldrich

Arsenic(III) oxide

SAJ first grade, ≥99.0%

Synonym(s):

Arsenic trioxide, Arsenous acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
As2O3
CAS Number:
Molecular Weight:
197.84
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
Pricing and availability is not currently available.

grade

SAJ first grade

vapor pressure

0.000001 hPa ( 66 °C)

Assay

≥99.0%

form

powder

availability

available only in Japan

SMILES string

O=[As]O[As]=O

InChI

1S/As2O3/c3-1-5-2-4

InChI key

IKWTVSLWAPBBKU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A - Eye Dam. 1 - Skin Corr. 1B - STOT RE 1

Target Organs

Respiratory system,Cardio-vascular system,Gastrointestinal tract

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

EU REACH Annex XIV (Authorisation List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yanfei Jia et al.
PloS one, 8(1), e54774-e54774 (2013-02-06)
Alpha-fetoprotein (AFP)-producing gastric cancer (AFPGC), represented by the production of AFP, has a more aggressive behavior than common gastric cancer. The underlying mechanisms are not well understood. Arsenic trioxide (As(2)O(3)) is used clinically to treat acute promyelocytic leukemia(APL) and has
Liangchun Hao et al.
Journal of pediatric hematology/oncology, 35(2), e67-e70 (2013-02-16)
The aim of this study was to investigate the hepatotoxicity induced by arsenic trioxide (As2O3) at a therapeutic dose for pediatric acute promyelocytic leukemia (APL). A total of APL patients received As2O3 treatment by IV drip. Hepatotoxicity was monitored based
Lauren A Held et al.
Cancer investigation, 31(3), 172-176 (2013-02-15)
This Phase I study assessed the feasibility of concomitant arsenic trioxide (ATO), ascorbic acid (AA), and bortezomib (Velcade™) (AAV) for patients with relapsed/refractory multiple myeloma. ATO (0.25 mg/kg) and AA (1 g) were given with an escalating dose of bortezomib
Angela M Mitchell et al.
Journal of virology, 87(8), 4571-4583 (2013-02-15)
Interactions with cellular stress pathways are central to the life cycle of many latent viruses. Here, we utilize adeno-associated virus (AAV) as a model to study these interactions, as previous studies have demonstrated that cellular stressors frequently increase transduction of
Hong Li et al.
Croatian medical journal, 54(1), 12-16 (2013-02-28)
To investigate the role of tumor apoptosis-inhibitory protein survivin in arsenic trioxide-induced apoptosis in VX-2 carcinoma in the rabbit liver by means of transcatheter arterial chemoembolization. Sixteen rabbits with 32 implanted hepatic VX-2 tumors were randomly divided into two groups.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service