Skip to Content
Merck

Skip To

NADPH-RO

Roche

NADPH

powder, =97% (dry weight)

Promotion Available

Synonym(s):

β-Nicotinamide adenine dinucleotide 2′-phosphate reduced tetrasodium salt hydrate, β-NADPH, 2′-NADPH hydrate, Coenzyme II reduced tetrasodium salt, Dihydronicotinamide adenine dinucleotide phosphate tetrasodium salt, NADPH Na4, TPNH2 Na4, Triphosphopyridine nucleotide reduced tetrasodium salt

Sign In to View Organizational & Contract Pricing.

Spend € 450 — Save 5 %; Spend € 700 — Save 10 %; Spend € 1.500 — Save 15 %*

Sign in to check eligibility. Shop Products

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mg
Check Cart for Availability
€ 297,00
500 mg
Check Cart for Availability
€ 1.260,00
1 g
Check Cart for Availability
€ 1.900,00

About This Item

Empirical Formula (Hill Notation):
C21H26N7Na4O17P3 · xH2O
CAS Number:
Molecular Weight:
833.35 (anhydrous basis)
UNSPSC Code:
41106305
PubChem Substance ID:
MDL number:
NACRES:
NA.54
Assay:
97% (dry weight)
Form:
powder
Storage temp.:
2-8°C

€ 297,00

Save Up To 15 %


Check Cart for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

Tetrasodium salt

assay

97% (dry weight)

form

powder

packaging

pkg of 1 g (10621706001), pkg of 100 mg (10107824001), pkg of 500 mg (10621692001)

manufacturer/tradename

Roche

λmax

260 and 340 nm

shipped in

wet ice

storage temp.

2-8°C

SMILES string

[Na+].[Na+].[Na+].[Na+].NC(=O)C1=CN(C=CC1)[C@H]2O[C@@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]3O[C@H]([C@H](OP([O-])([O-])=O)[C@@H]3O)n4cnc5c(N)ncnc45)[C@H](O)[C@@H]2O

InChI

1S/C21H30N7O17P3.4Na/c22-17-12-19(25-7-24-17)28(8-26-12)21-16(44-46(33,34)35)14(30)11(43-21)6-41-48(38,39)45-47(36,37)40-5-10-13(29)15(31)20(42-10)27-3-1-2-9(4-27)18(23)32;;;;/h1,3-4,7-8,10-11,13-16,20-21,29-31H,2,5-6H2,(H2,23,32)(H,36,37)(H,38,39)(H2,22,24,25)(H2,33,34,35);;;;/q;4*+1/p-4/t10-,11+,13-,14+,15-,16+,20-,21+;;;;/m0..../s1

InChI key

WYWWVJHQDVCHKF-MPUNMZHWSA-J

General description

  • Absorbance coefficients: 15.0 x 103 l x mol-1 x cm-1 at 260 nm, 6.3 x 103 l x mol-1 x cm-1 at 340 nm
  • Absorbance maxima: 260 nm and 340 nm
  • Molecular weight: NADPH-Na4: Mr = 833.4, NADPH: Mr = 745.4
  • pH value in water: 8.0 ± 1.0 (10 mg/ml)
Amorphous powder bottled under nitrogen.

Application

NADPH-RO has been used for the determination of thioredoxin reductase activity.

Biochem/physiol Actions

NADPH is an electron donor and a cofactor for many redox enzymes including nitric oxide synthetase.

Preparation Note

Storage conditions (working solution): 2 to 8 °C
10 mg/ml in Na-bicarbonate buffer, pH 9, contains at least 95% NADPH after 4 weeks at 2 to 8 °C.

Analysis Note

  • Purity: =97% β-NADPH-Na4 based on dry weight (enzymatically and absorbance at 340 nm), =80% (absorbance at 260 nm)
  • Organic solvents: =3% (GC)
Absorbance coefficients: 15.0 x 103 l x mol-1 x cm-1 at 260 nm, 6.3 x 103 l x mol-1 x cm-1 at 340 nm

Other Notes

For life science research only. Not for use in diagnostic procedures.
Packaged based on solid weight.


Storage Class

11 - Combustible Solids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library