Skip to Content
Merck
All Photos(1)

Key Documents

G6PDHII-RO

Roche

Glucose-6-Phosphate Dehydrogenase (G6P-DH)

grade II, from yeast

Synonym(s):

G6P-DH, Glucose-6-Phosephate Dehydrogenase

Sign Into View Organizational & Contract Pricing

Select a Size

2 ML
€277.00

€277.00


Check Cart for Availability


Select a Size

Change View
2 ML
€277.00

About This Item

Enzyme Commission number:
UNSPSC Code:
12352204

€277.00


Check Cart for Availability

biological source

yeast

Quality Level

form

suspension

specific activity

~140 units/mg protein (At 25 °C with glucose-6-P as the substrate; standardized with BSA.)

packaging

pkg of 2 mL (10127671001 [5 mg/mL])
pkg of 5 mL (10737232001 [5 mg/mL])

manufacturer/tradename

Roche

optimum pH

9.2

pH range

5-11

shipped in

wet ice

General description

Glucose-6-Phosphate Dehydrogenase (G6PD) is a ubiquitous enzyme which acts as a catalyst in producing pentose. It also produces NADPH for various biosynthetic and detoxification reactions. Sometimes G6PD provides an alternative to main glycolytic pathway for glucose utilization. It also has its application as DNA markers on X-chromosome.

Application

Glucose-6-Phosphate Dehydrogenase (G6P-DH) is a component of cofactor recycling systems for NADPH. Additional reagent glucose-6-phosphate.
It was used in enzymatic determination of C6 phosphorylation of glycogen.[1]

Quality

Contaminants: <0.001% CK, <0.05% GR, <0.01% 6-PGDH and PGluM each, <0.02% HK, <0.002% PGI

Physical form

Suspension in 3.2 M ammonium sulfate solution, pH approximately 6.0

Preparation Note

Activator: Mg2+ (5 to 10 mM)

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna A DePaoli-Roach et al.
The Journal of biological chemistry, 290(2), 841-850 (2014-11-25)
Glycogen is a branched polymer of glucose that acts as an energy reserve in many cell types. Glycogen contains trace amounts of covalent phosphate, in the range of 1 phosphate per 500-2000 glucose residues depending on the source. The function

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service