Skip to Content
Merck
All Photos(2)

Key Documents

12015196001

Roche

Lumi-LightPLUS Western Blotting Substrate

pkg of 100 mL (1,000 cm2 membrane)

Synonym(s):

western blotting substrate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
Pricing and availability is not currently available.

packaging

pkg of 100 mL (1,000 cm2 membrane)

Quality Level

manufacturer/tradename

Roche

shipped in

wet ice

storage temp.

2-8°C

General description

As the technology for quantifying chemiluminescence advances, the substrates for chemiluminescence must also advance. For instance, the light emission of conventional enhanced luminol substrates substantially decreases within 30 minutes. In contrast, the chemiluminescence signal of the Lumi-LightPLUS substrate persists for more than nine hours, and since exposures of only a few minutes are necessary, multiple exposures can be taken. Additionally, the ability to take extended exposures allows the detection of rare proteins, or higher dilutions of antigen or antibody to be used. The emission wavelength of the Lumi-LightPLUS substrate is 425 nm.

Comparison of Lumi-Light Substrate and Lumi-Light PLUS Substrate

Lumi-Light Substrate Lumi-LightPLUS Substrate
Sensitivity
10-50 pg antigen 1-5 pg antigen
Pimary Antibody Dilution Dilute up to 10-fold more compared to colorimetric detection Dilute up to 100-fold more compared to colorimetric detection
Signal Stability >3 hours (enhance signal with a 30-minute incubation) > 9 hours (enhance signal with a 30-minute incubation)

Number of tests: The reagent is sufficient for 10 blots, each with a size of 10 cm x 10 cm.
Sensitivity: Depending on the affinity of the primary antibody, 1 - 5 pg of antigen can be detected in research studies.
Storage/stability: The Lumi-LightPLUS substrate components are stable at +2 to +8°C until the expiration date printed on the label. Components are shipped at +15 to +25°C.

Application

Lumi-LightPLUS Western Blotting Substrate has been used in western blotting.[1][2][3]
Lumi-LightPLUS Western Blotting Substrate is for the chemiluminescent detection of antigens blotted on membranes using horseradish peroxidase-conjugated secondary antibodies. It can be used for very sensitive detection of any antigen blotted on a PVDF Western Blotting Membrane or nitrocellulose membrane.

Lumi-LightPLUS Western Blotting Substrate is suited for high-sensitivity western blotting, especially when quantification is required. The chemiluminescent signal persists for more than 9 hours, allowing multiple exposures to be taken.

Features and Benefits

  • Detect rare proteins. Detect antigen in the range of 1 - 5 pg.
  • Take multiple exposures. The signal is stable for more than nine hours after substrate addition.
  • Save sample material. Lumi-LightPLUS substrate detects less than 5 pg-amounts of antigen.
  • Save valuable resources. Dilute primary antibody 10- to 100-fold more than with colorimetric detection systems.
  • Benefit from easy preparation. Simply mix the two Lumi-Light components in a 1:1 ratio.

Packaging

1 kit containing 2 components

Quality

Each lot is function tested using mouse or rabbit primary antibodies.

Preparation Note

Stabilizers: proteinaceous stabilizers
Working concentration: The working concentration of conjugate depends on application and substrate.
The following concentrations should be taken as a guideline:
  • Dot blot: 50 mU/ml
  • ELISA: 25 mU/ml
  • Western blot: 50 mU/ml


Working solution: Preparation of Additional Reagents and Solutions

The table inside the package insert describes the preparation of working solutions. Volumes are designed for a membrane of 10x10 cm. If larger membranes are used, volumes have to be scaled up. For reproducible results, equilibrate all solutions to room temperature before use.
Note: Do not use Azide to stabilize the solutions against microbial growth, as azide irreversibly inhibits horseradish peroxidase.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Kit Components Only

Product No.
Description

  • Lumi-LightPLUS Luminol/Enhancer Solution

  • Lumi-LightPLUS Stable Peroxide Solution

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Serum-Induced Differentiation of Glioblastoma Neurospheres Leads to Enhanced Migration/Invasion Capacity That Is Associated with Increased MMP9
Joseph JV, et al.
PLoS ONE (2015)
Ataxia telangiectasia mutated (ATM) inhibition transforms human mammary gland epithelial cells
Mandriota SJ, et al.
The Journal of Biological Chemistry, 285(17), 13092-13106 (2010)
Hana Cernecka et al.
Naunyn-Schmiedeberg's archives of pharmacology, 385(9), 875-882 (2012-06-13)
β(3)-Adrenoceptors are a promising drug target for the treatment of urinary bladder dysfunction, but knowledge about their expression at the protein level and their functional role is limited, partly due to a lack of well validated tools. As many antibodies
Maria Samata et al.
Cell, 182(1), 127-144 (2020-06-06)
Before zygotic genome activation (ZGA), the quiescent genome undergoes reprogramming to transition into the transcriptionally active state. However, the mechanisms underlying euchromatin establishment during early embryogenesis remain poorly understood. Here, we show that histone H4 lysine 16 acetylation (H4K16ac) is
Thioredoxin-1 protects against androgen receptor-induced redox vulnerability in castration-resistant prostate cancer
Samaranayake GJ, et al.
Nature Communications, 8(1), 1204-1204 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service