Skip to Content
Merck
All Photos(1)

Documents

11666690001

Roche

Buffers in a Box, Premixed TAE Buffer, 10x

pH 8.4, suitable for electrophoresis, pkg of 4 L, solution

Synonym(s):

TAE buffer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

form

solution

packaging

pkg of 4 L

manufacturer/tradename

Roche

technique(s)

electrophoresis: suitable

pH

8.4

shipped in

ambient

storage temp.

20-25°C

General description

10x solution

Quality

Absence of contaminants: Free of DNase and RNase according to the current Quality Control procedures.

Physical form

Appearance: clear, colorless solution

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lindsey A Lonowski et al.
Nature protocols, 12(3), 581-603 (2017-02-17)
This protocol describes methods for increasing and evaluating the efficiency of genome editing based on the CRISPR-Cas9 (clustered regularly interspaced short palindromic repeats-CRISPR-associated 9) system, transcription activator-like effector nucleases (TALENs) or zinc-finger nucleases (ZFNs). First, Indel Detection by Amplicon Analysis
Brian A Sanderson et al.
Analytical biochemistry, 454, 44-52 (2014-03-19)
Agarose gel electrophoresis of DNA and RNA is routinely performed using buffers containing either Tris, acetate, and EDTA (TAE) or Tris, borate, and EDTA (TBE). Gels are run at a low, constant voltage (∼10 V/cm) to minimize current and asymmetric

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service