Skip to Content
Merck
All Photos(1)

Key Documents

11426320001

Roche

Anti-Fluorescein

from mouse IgG1 (clone B13-DE1)

Synonym(s):

antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

B13-DE1, monoclonal

form

lyophilized

packaging

pkg of 100 μg

manufacturer/tradename

Roche

isotype

IgG1

storage temp.

2-8°C

General description

Monoclonal antibody to fluorescein.
The detection of bound antibody can be carried out directly in one step using an anti-mouse Ig fluorochrome/enzyme conjugate, or in a two-step procedure with anti-mouse Ig fluorescein and, subsequently, anti-fluorescein enzyme conjugate.
The antibody does not contain any protein and can hence be used for coating and labeling.

Contents
Lyophilizate, stabilized

Specificity

The monoclonal antibody reacts with free and bound fluorescein.

Application

Use Anti-Fluorescein for the detection of fluorescein-labeled compounds using:
  • ELISA
  • Immunohistocytochemistry
  • In situ hybridization
  • Western blot
Anti-Fluorescein has been used to capture amplicon in quadruple-tagging PCR and nucleic acid lateral flow assay for simultaneous detection of foodborne pathogens.

Preparation Note

Working concentration: Working concentration of conjugate depends on application and substrate. The following concentrations should be taken as a guideline:
  • ELISA: 2 to 4 μg/ml
  • Immunohistocytochemistry: 0.5 to 2 μg/ml
  • In situ hybridization: 0.2 to 0.4 μg/ml
  • Western blot: 0.5 to 2 μg/ml

Working solution: Phosphate buffered saline, pH 7.4
Using water instead the antibody can precipitate.

Reconstitution

Add 1 ml double-distilled water to a final concentration of 100 μg/ml.
Reconstitute for 15 minutes.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ignacio E Schor et al.
Current biology : CB, 28(22), 3547-3561 (2018-11-06)
Long non-coding RNAs (lncRNAs) can often function in the regulation of gene expression during development; however, their generality as essential regulators in developmental processes and organismal phenotypes remains unclear. Here, we performed a tailored investigation of lncRNA expression and function
J Spielmann et al.
BMC obesity, 4, 24-24 (2017-07-12)
Obesity was identified as a major risk factor for malignant diseases, but underlying mechanisms remain unclear. Natural killer (NK) cells, a pivotal aspect of innate immunity, are capable of identifying and killing virally infected and tumor cells. Previous studies have
Comparing nucleic acid lateral flow and electrochemical genosensing for the simultaneous detection of foodborne pathogens.
Aissa A B, et al.
Biosensors And Bioelectronics, 88, 265-272 (2017)
Yufeng Liu et al.
Oncology reports, 44(2), 499-508 (2020-07-07)
Apurinic/apyrimidinic endonuclease 1 (APE1) is a primary nuclear‑localized multifunctional protein in osteosarcoma. However, the cytoplasmic localization of APE1 was found to be functional and to increase with cisplatin resistance, yet the molecular mechanism is unknown. In the present study, we explored
María Fernanda Prado-Fernández et al.
Current issues in molecular biology, 46(7), 6885-6902 (2024-07-26)
Pereskia sacharosa Griseb. is a plant used in traditional herbal medicine to treat inflammation. We analyzed the phenolic content of P. sacharosa leaves (EEPs) by liquid chromatography-tandem mass spectrometry (LC-MS/MS) and investigated the anti-inflammatory properties of EEPs and its flavonoid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service