Skip to Content
Merck
All Photos(1)

Key Documents

10904988001

Roche

m7G(5′)ppp(5′)G

P1-5′-(7-Methyl)-guanosine-P3-5′-guanosine triphosphate

Synonym(s):

methyl-guanosine-p3-5′-guanosine-triphosphate, p1-5′-7-, p1-5′-(7-methyl)-guanosine-p3-5′-guanosine-triphosphate | methyl-guanosine-p3-5′-guanosine-triphosphate, p1-5′-7-

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116100

form

lyophilized

mol wt

815.3

packaging

pkg of 5 U

manufacturer/tradename

Roche

storage temp.

−20°C

Application

The nucleotide is used in combination with the SP6 and T7 RNA polymerase promoter system as an initiator for in vitro RNA synthesis.

Quality

Typical analysis: 90% m7G(5′)ppp(5′)G (from Porg.), 1.7% lithium

Specifications

Formula: C21H28N10O18P3Li2

Unit Definition

Conversion Factor
The data for converting A250 units into mg is: 5 A250 units = 0.2347 μM = 0.1914 mg.

Physical form

Lyophilizate

Preparation Note

Working concentration: The recommended working concentration with T7, T3 or SP6 RNA Polymerases is 1 mM.

Reconstitution

For best results, dissolve in water or TE at 10 mM.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Understand how mRNA vaccines induce immunity. and read how synthetic mRNA is prepared for vaccine immunogens and other biopharmaceuticals. Find reagents for synthesis of mRNA.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service