Skip to Content
Merck
All Photos(1)

Key Documents

10107085001

Roche

L-Lactate Dehydrogenase (L-LDH)

from hog muscle

Synonym(s):

L-LDH, L-Lactate Dehydrogenase, L-lactate

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
12352204
Pricing and availability is not currently available.

biological source

hog muscle

Quality Level

form

suspension

specific activity

~550 units/mg protein (at 25 °C (1,000 U/mg at 37 °C) with pyruvate as the substrate.)

packaging

pkg of 10 mL (100 mg)

manufacturer/tradename

Roche

Related Categories

General description

L-Lactate Dehydrogenase (L-LDH) is an enzyme which is involved in the glycolytic pathway. It is believed to consist of five isotypes, LDH1 to LDH5.[1] It also has a major role in carbohydrate metabolism of human malaria parasites.[2]

Application

It was used in determination of D-Lactate.[3]
Reduction of α-ketoacids to α-hydroxycarboxylic acids or reverse reaction.

Physical form

Suspension in 3.2 M ammonium sulfate solution, pH approximately 6.5

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mizeck Gg Chagunda et al.
The Journal of dairy research, 73(4), 431-440 (2006-07-11)
Systematic factors affecting the activities of L-lactate dehydrogenase (LDH) and N-acetyl-beta-D-glucosaminidase (NAGase) and somatic cell count (SCC), the association between the activities of LDH and NAGase and SCC with respect to udder health status, and the ability of LDH and
Plasmodium falciparum and Plasmodium vivax: Lactate-Dehydrogenase Activity and Its Application for in Vitro Drug Susceptibility Assay.
Basco, Leonardo K., et al.
Experimental Parasitology, 80.2, 260-271 (1995)
Thorsten Irsch et al.
The Journal of biological chemistry, 279(21), 22209-22217 (2004-02-21)
The glyoxalase system is a ubiquitous pathway catalyzing the glutathione-dependent detoxication of ketoaldehydes such as methylglyoxal, which is mainly formed as a by-product of glycolysis. The gene encoding a glyoxalase II has been cloned from Trypanosoma brucei, the causative agent
Heiko Henning et al.
Andrology, 10(7), 1426-1440 (2022-07-06)
Prolonging the shelf-life of liquid-preserved semen without compromising its fertilizing capacity may increase the efficiency of artificial insemination in pigs. Many fertilization-relevant processes are adenosine triphosphate dependent. The impact of semen storage and rewarming to body temperature on the energy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service