05480647001
Roche
COT Human DNA, Fluorometric Grade
solution, pkg of 1 mg
Synonym(s):
COT Human DNA, DNA
Select a Size
€628.00
Select a Size
About This Item
€628.00
Recommended Products
biological source
human placenta (DNA)
Quality Level
form
solution
packaging
pkg of 1 mg
manufacturer/tradename
Roche
concentration
1.0-1.5 mg/mL
impurities
HCV/HBV, none detected
HIV 1/2, none detected
color
colorless
solubility
water: miscible
storage temp.
−20°C (−15°C to −25°C)
Looking for similar products? Visit Product Comparison Guide
General description
Application
- Nucleic acid labeling and detection
- DNA Microarray applications such as comparative genome hybridization and sequence capture
- Complex hybridization of human nucleic acids like FISH
Features and Benefits
- Improve specificity by blocking unspecific nucleic acid motifs using high concentrations of double-stranded, repetitive, human DNA.
- Achieve consistent and reliable results using fluorometrically quantified COT Human DNA, for preferentially detecting dsDNA.
- Use COT Human DNA that has been tested using DNA microarrays and comparative genome hybridization (CGH).
Quality
For quality assessment in the CGH application, target sequences located on the X-chromosome are compared in male and female genomic DNA samples. The donor material for this product was tested for HBs antigen and for the presence of antibodies to HIV-1, HIV-2, HCV, and found to be negative.
Sequence
Physical form
Storage and Stability
Other Notes
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
No data available
Flash Point(C)
No data available
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service