RIE213109881
Rieke Metals, Inc.
2-Fluorophenethylmagnesium bromide 0.5 M in Tetrahydrofuran
0.5 M in Tetrahydrofuran
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Molecular Weight:
227.36
MDL number:
Iupac Name
bromo[2-(2-fluorophenyl)ethyl]magnesium
Inchi Code
1S/C8H8F.BrH.Mg/c1-2-7-5-3-4-6-8(7)9;;/h3-6H,1-2H2;1H;/q;;+1/p-1
InChI key
WEIRNJAUMBDPAY-UHFFFAOYSA-M
Country of Origin
US
Physical Form
Liquid
Product Link
https://riekemetals.com/products-services/catnum/3233
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.