ZSTWIN0A1
Tank Level Adapter
Transmits tank pure water feeding level information to Milli-Q® IQ 7000 and EQ 7000 systems
All Photos(1)
About This Item
Recommended Products
packaging
pkg of 1 unit
compatibility
for use with Milli-Q® EQ 7000
for use with Milli-Q® IQ 7000
General description
This electronic sensor transfers tank level information to the Milli-Q® IQ 7000 and Milli-Q® EQ 7000 water purification systems. It stops water feed from an empty tank and lets you check water levels in the tank before sourcing water, preventing air from entering the system.
Application
Transmits the polyethylene (PE) tank pure water feeding level information to Milli-Q® IQ 7000 and Milli-Q® EQ 7000 ultrapure water purification systems.
Legal Information
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany