Skip to Content
Merck
All Photos(1)

Key Documents

XX1102012

Millipore

Plastic Syringes, 20mL Luer slip, polypropylene

Beer, Bottled Water, Cosmetics, Pharmaceutical Products Analysis, Bioburden Testing

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NB.11
Pricing and availability is not currently available.

material

clear device
polypropylene device

Quality Level

sterility

non-sterile

parameter

20 mL sample volume

input

sample type bottled water
sample type cosmetics
sample type beer
sample type pharmaceutical(s)

application(s)

bioburden testing
cosmetics
pharmaceutical
water monitoring

shipped in

ambient

General description

Use polypropylene syringes to dispense liquids into small containers and pressure-filter small volumes through filter holders or filter units with female Luer slip inlets

Application

Beer, Bottled Water, Cosmetics, Pharmaceutical Products Analysis, Bioburden Testing, General microbiological analysis.

Specific test regulations for general microbiological analysis.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Photometric determination of phosphate ions in water with ammonium molybdate using Spectroquant® test kits and a UV/VIS spectrophotometer.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service