Skip to Content
Merck
All Photos(1)

Key Documents

WMEQ0RKT

Milli-Q

Milli-Q® EQ Wall Mounting Kit

Wall mounting kit for the Q-POD® of Milli-Q® EQ series water purification systems.

Synonym(s):

Milli-Q® EQ wall mounting kit for POD

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
€1,330.00

€1,330.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 EACH
€1,330.00

About This Item

UNSPSC Code:
41104200
NACRES:
JA.13

€1,330.00


Check Cart for Availability

Request more information

packaging

pkg of 1 unit

compatibility

for use with Milli-Q® EQ 7000
for use with Milli-Q® EQ 7008
for use with Milli-Q® EQ 7016

Looking for similar products? Visit Product Comparison Guide

General description

The wall mounting kit is a water system accessory that includes all necessary components for convenient installation of the Q-POD® dispenser of Milli-Q® EQ 7000/08/16 water purification systems. The dispenser can be wall-mounted up to 3 m from the system, saving space.
This kit optimizes the installation flexibility of the system. If your lab changes, the system set-up can adapt to your lab.

Features and Benefits

  • Saves lab space.
  • Ease of installation.

Legal Information

Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
Q-POD is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service