Skip to Content
Merck

Skip To

VMWP02500

MCE Membrane Filter, 0.05 μm Pore Size

greener alternative

MF-Millipore, filter diam. 25 mm, hydrophilic

Synonym(s):

MF-Millipore Membrane Filter, 0.05 µm pore size

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
40161507
NACRES:
NB.24
eCl@ss:
32031602
Pore size:
0.05 μm pore size, 72 % porosity
Material:
mixed cellulose esters (MCE) membrane, plain filter, white filter
Feature:
hydrophilic
Diam.:
25 mm
Sterility:
non-sterile
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

material

mixed cellulose esters (MCE) membrane, plain filter, white filter

description

25 mm diameter, mixed cellulose esters (MCE) membrane, hydrophilic, white, 100 discs

sterility

non-sterile

feature

hydrophilic

manufacturer/tradename

MF-Millipore, Millipore

sustainability

Greener Alternative Product

parameter

0.25 L/min-cm2 air flow rate, 75 °C max. temp., 0.74 mL/min-cm2 water flow rate

diam.

25 mm

filter diam.

25 mm

thickness

100 μm

gravimetric extractables

1.5%

refractive index

n/D 1.5

matrix

MF-Millipore

pore size

0.05 μm pore size, 72 % porosity

bubble point

≥17.6 bar, air with water at 23 °C

greener alternative category

shipped in

ambient

General description

We are committed to bringing you Greener Alternative Products, which have been re-engineered to enable more sustainable research solutions. The inner & outer boxes containing the membrane discs are made from bio-circular material allocated using the ISCC mass balance approach. Click here for more information.

Application

Microdialysis of DNA and proteins

Other Notes

Directions for Use

  • Organism Retention: Microorganism
  • Mode of Action: Filtration (size exclusion)
  • Application: General laboratory filtration
  • Intended Use: Retention or removal of biological contaminants
  • Instructions for Use: Sterilizing filtration of a liquid through a membrane with a 0.2 μm (or smaller) pore size effectively removes biological contaminants, including bacteria, mold and yeast. For the selective retention of larger biological contaminents, liquid filtration through membranes with 0.45 μm (or larger) pore sizes may be used to trap and support microorganism growth for subsequent culture and analysis
  • Storage Statement: Store in dry location away from heat source
  • Disposal Statement: Dispose of in accordance with applicable federal, state and local regulations.

Legal Information

MF-Millipore is a trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
VMWP04700VMWP01300VMWP09025
pore size

0.05 μm pore size, 72 % porosity

pore size

0.05 μm pore size, 72 % porosity

pore size

0.05 μm pore size, 72 % porosity

pore size

0.05 μm pore size, 72 % porosity

manufacturer/tradename

MF-Millipore

manufacturer/tradename

MF-Millipore

manufacturer/tradename

MF-Millipore

manufacturer/tradename

MF-Millipore

material

mixed cellulose esters (MCE) membrane, white filter, plain filter

material

mixed cellulose esters (MCE) membrane, plain filter, white filter

material

mixed cellulose esters (MCE) membrane, plain filter, white filter

material

mixed cellulose esters (MCE) membrane, plain filter, white filter

diam.

25 mm

diam.

47 mm

diam.

13 mm

diam.

90 mm

parameter

0.25 L/min-cm2 air flow rate, 75 °C max. temp., 0.74 mL/min-cm2 water flow rate

parameter

0.25 L/min-cm2 air flow rate, 0.74 mL/min-cm2 water flow rate, 75 °C max. temp.

parameter

0.25 L/min-cm2 air flow rate, 0.74 mL/min-cm2 water flow rate, 75 °C max. temp.

parameter

0.25 L/min-cm2 air flow rate, 0.74 mL/min-cm2 water flow rate, 75 °C max. temp.

sterility

non-sterile

sterility

non-sterile

sterility

non-sterile

sterility

non-sterile


Still not finding the right product?


signalword

Danger

Storage Class

4.1B - Flammable solid hazardous materials

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - ED ENV 1 - Eye Irrit. 2 - Flam. Sol. 1


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

9036-19-5

CAS No.

9036-19-5

CAS No.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters