TP-10L-BK
Bulk Reagent - ABO & RHD (Rh) Specificities
BIOSCOT™ Anti-D from the clone MS-201, Human IgM, Blood Grouping Reagent
Synonym(s):
Anti-D (clone MS-201), Blood Grouping Reagent
About This Item
Recommended Products
biological source
human
Quality Level
antibody form
unpurified
clone
MS-201, monoclonal
form
liquid
packaging
pkg of 10 L
concentration
40-60 mg/mL protein (Total Protein Range)
technique(s)
agglutination assay: suitable
pH
7.0-7.4
isotype
IgM
shipped in
ambient
storage temp.
2-8°C
Related Categories
General description
Contains 0.1% (w/v) Sodium Azide
Filtered through a 0.22μm terminal filter
Shelf Life: 36 months from date of filtration
Product formulation examined according to Annex IV of the EU IVD directive to support CE registration.
For In Vitro use only.
Application
Legal Information
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service