Skip to Content
Merck

TK-5ML-B

Blood typing BIOSCOT IVD Reagent RHCE (Rh) Specificities

BIOSCOT Anti-c from the clone MS-33, Human IgM, Blood Grouping Reagent

Synonym(s):

Anti-c (clone MS-33), Blood Grouping Reagent

Sign In to View Organizational & Contract Pricing.
Note: For further information on price, availability or to place an order, please contact us

About This Item

NACRES:
NA.41
UNSPSC Code:
41116102
Clone:
MS-33, monoclonal
Technique(s):
agglutination assay: suitable
Application:
Agglut.
Citations:
-

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

human

clone

MS-33, monoclonal

packaging

pkg of 5 mL

manufacturer/tradename

BIOSCOT

storage condition

do not freeze

concentration

45-60 mg/mL protein (Total Protein Range)

technique(s)

agglutination assay: suitable

pH

7.1-7.3

isotype

IgM

application(s)

clinical testing

shipped in

ambient

storage temp.

2-8°C

Quality Level

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
TK-1L-BKTK-10X5ML-BTK-10X5ML-U
clone

MS-33, monoclonal

clone

MS-33, monoclonal

clone

MS-33, monoclonal

clone

MS-33, monoclonal

biological source

human

biological source

human

biological source

human

biological source

human

technique(s)

agglutination assay: suitable

technique(s)

agglutination assay: suitable

technique(s)

agglutination assay: suitable

technique(s)

agglutination assay: suitable

Quality Level

600

Quality Level

600

Quality Level

600

Quality Level

600

isotype

IgM

isotype

IgM

isotype

IgM

isotype

IgM

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

ambient

General description

Contains 0.1% (w/v) Sodium Azide

Filtered through a 0.22μm terminal filter

Shelf Life: 36 months from date of filtration

CE marked under the requirements of EU regulation 2017/746




For In Vitro use only .

Application

For use by Tube;Slide;Microplate;Gel and Column techniques
Vialled, Labeled

Legal Information

Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
BIOSCOT is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service